Analytical Acetone
Acetone, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Isopropanol Acetonitrile Acetone (9/9/2) Flush Solution, Fisher Chemical
Chemical Name or Material: 2-Propanol (45%), Acetonitrile (45%), and Acetone (10%) Solution Name Note: Mobile-phase solvent blend CAS: 67-63-0 CAS: 67-64-1 CAS: 75-05-8 Physical Form: Liquid Assay: Pass Test (By GC-FID)
Acetone, >95%, technical, ACROS Organics™
Chemical Name or Material: Acetone Molecular Formula: C3H6O Formula Weight: 58.08 Synonym: 2-Propanone
Acetone, ACS, 99.5+%, Alfa Aesar
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: ACS Reagent Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765
Acetone (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: Assay: ≥99.5% Molecular Formula: C3H6O Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.6%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: Reagent ACS, Spectro Grade CAS: 67-64-1 CAS Min %: 99.5 CAS Max %: 100.0 Purity Grade: ACS Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone Products for Science Education
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: 100% v/v Linear Formula: CH3COCH3 Formula Weight: 58.08 Synonym: 2-Propanone
Acetone, 99.5+%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: For analysis CAS: 67-64-1 CAS Min %: 99.5 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.50% Assay Percent Range: 99.5+% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: Water <50 ppm CAS: 67-64-1 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 CAS Min %: 99.5 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (ACS AR), Macron Fine Chemicals™
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: ACS Reagent Grade Molecular Formula: CH3COCH3 Formula Weight: 58.08
Acetone/Acetonitrile 80/20, (HPLC), Ricca Chemical
Chemical Name or Material: Acetone/Acetonitrile Name Note: 80 (Acetone) / 20 (Acetonitrile) CAS: 67-64-1 CAS Min %: 78.0 CAS Max %: 82.0 CAS: 75-05-8 CAS Min %: 19.59 CAS Max %: 20.39 Purity Grade: HPLC Grade Physical Form: Liquid
Acetone, HPLC Grade, Ricca Chemical
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: HPLC Grade
Acetone, AR, For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Acetone Name Note: ACS CAS: 67-64-1 Physical Form: Liquid Molecular Formula: CH3COCH3 Formula Weight: 58.08
Acetone (ACS), Ricca Chemical
Chemical Name or Material: Acetone CAS: 67-64-1 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade
Acetone, 90% (v/v) Aqueous, Ricca Chemical
Chemical Name or Material: Acetone CAS: 67-64-1 CAS Min %: 86.0 CAS Max %: 89.0 CAS: 7732-18-5 CAS Min %: 12.11 CAS Max %: 12.61 Physical Form: Liquid
Acetone (Buffered TS), Ricca Chemical
Chemical Name or Material: Acetone, Buffered Name Note: Test Solution CAS: 7732-18-5 CAS Min %: 76.0 CAS Max %: 79.0 CAS: 67-64-1 CAS Min %: 15.73 CAS Max %: 16.37 CAS: 7647-14-5 CAS Min %: 5.59 CAS Max %: 5.82 CAS: 6131-90-4 CAS Min %: 1.09 CAS Max %: 1.13 CAS: 7647-01-0 CAS Min %: 0.09 CAS Max %: 0.09 Physical Form: Liquid
Acetone, Macron Fine Chemicals™
Chemical Name or Material: Acetone CAS: 67-64-1 Formula Weight: 58.08
Acetone, ACS Reagent Grade ≥99.5%, MP Biomedicals™
Chemical Name or Material: Acetone Name Note: For ACS analysis CAS: 67-64-1 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: ≥99.5% Molecular Formula: C3H6O Formula Weight: 58.1
Acetone, 99.5% min, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: For ACS analysis CAS: 67-64-1 Purity Grade: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs Physical Form: Liquid Assay: ≥99.5% (GC, corrected for water content) Assay Percent Range: 99.5% min. Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 Synonym: 2- Propanone
Acetone, ≥95%, Ultra Scientific
Chemical Name or Material: Acetone CAS: 000067-64-1 CAS Max %: 100.0 Assay Percent Range: >95%
  spinner