Analytical Acetonitrile
Acetonitrile with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade Available on GSA/VA Contract for Federal Government customers only.
Aluminum (Al): 25 ppb max. Assay Percent Range: 40 to 42 % Calcium (Ca): 50 ppb max. CAS: 64-18-6 CAS: 64-18-6 Chemical Name or Material: 0.1% Formic Acid in Acetonitrile (v/v) Copper (Cu): 10 ppb max. Density: 0.78g/mL Color: Clear Formula Weight: 41.05 Physical Form: Liquid Purity Grade: Optima™ LC/MS DOT Information: DOT Class 3, : Flammable Liquid Formic Acid: 0.095 to 0.105% Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0 Identification: Pass Test Iron (Fe): 10 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 10 ppb max. Linear Formula: CH3CN Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 4269 MDL Number: 3297 Molecular Formula: CH2O2 Name Note: Solvent Blends Nickel (Ni): 10 ppb max. Packaging: Glass Bottle Potassium (K): 20 ppb max. Residue after Evaporation: 1ppm max. Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Water: 0.01% max. Zinc (Zn): 20 ppb max.
Acetonitrile with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade
Aluminum (Al): 25 ppb max. Assay Percent Range: ≥97.5 % Calcium (Ca): 50 ppb max. CAS: 76-05-1 CAS: 75-05-8 Chemical Name or Material: 0.1% Trifluoroacetic Acid in Acetonitrile (v/v) Copper (Cu): 10 ppb max. Decomposition Information: >500°C Density: 1.42 (Air = 1.0) Assay: 0.095 to 0.105% (v/v) Trifluoroacetic Acid Boiling Point: 82°C Color: Clear Formula Weight: 41.04 Melting Point: −50°C pH: 2 Physical Form: Liquid Purity Grade: Optima™ LC/MS DOT Information: DOT Class 3, : Flammable Liquid Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:0 Identification: Pass Test Impurities: ≤20ppb (Ionic, Potassium) Iron (Fe): 10 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 10 ppb max. Linear Formula: CH3CN Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 9847 MDL Number: 4169 Molecular Formula: C2HF3O2 Name Note: Solvent Blends Nickel (Ni): 10 ppb max. Packaging: Glass Bottle Potassium (K): 20 ppb max. Residue after Evaporation: ≤1ppm Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Trifluoroacetic Acid: Inclusive between 0.095% Vapor Density: 1.42 Water: ≤0.01% Zinc (Zn): 20 ppb max.
Acetonitrile with trifluoroacetic acid (0.05%), Mobile phase for HPLC, Fisher Chemical
Chemical Name or Material: Acetonitrile with 0.05% Trifluoroacetic Acid (v/v) CAS: 76-05-1 CAS: 75-05-8 Purity Grade: HPLC Grade Assay: Inclusive between 0.0475% MDL Number: MFCD00001878
Acetonitrile, HPLC Gradient Grade, ACROS Organics™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC Gradient Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: AN
Mobile phase for HPLC, Acetonitrile with trifluoroacetic acid (0.1%), HPLC Grade, Fisher Chemical
Chemical Name or Material: Acetonitrile with 0.1% Trifluoroacetic Acid (v/v) CAS: 76-05-1 CAS: 75-05-8 Purity Grade: HPLC Grade Purity Grade Notes: Blended mobile phase for HPLC applications Assay: Inclusive between 0.095% Molecular Formula: C2H3N Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: 0.1% TFA in ACN
Acetonitrile, anhydrous, >99.8%, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile (Certified ACS), Fisher Chemical
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Ethanenitrile
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: For analysis ACS CAS: 75-05-8 CAS Min %: 99.5 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, Anhydrous, Amine free, 99.9%, Alfa Aesar™
Chemical Name or Material: Acetonitrile Name Note: Amine Free CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, Environmental Grade, 99.7+%, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, anhydrous, >99.8%, Alfa Aesar
Chemical Name or Material: Acetonitrile, anhydrous Name Note: Water <100ppm CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878
Acetonitrile, 99.9+%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.90% (GC) Assay Percent Range: 99.9+% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: water <50 ppm, extra dry over mol. sieve CAS: 75-05-8 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.85% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Mobile phase for HPLC, Acetonitrile with formic acid (0.1%), HPLC Grade, Fisher Chemical
Chemical Name or Material: Acetonitrile with 0.1% Formic Acid (v/v) CAS: 75-05-8 CAS: 64-18-6 Purity Grade: HPLC Grade Assay Percent Range: ≥98.5 % MDL Number: MFCD00001878
Acetonitrile, Supergradient HPLC Grade (Far UV), 99.9+%, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.9% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Extra Dry Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, ACS, >99.5%, Alfa Aesar™
Chemical Name or Material: Acetonitrile Name Note: For analysis ACS CAS: 75-05-8 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Acetonitrile Name Note: AcroSeal™, Quadrant Cap CAS: 75-05-8 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile Anhydrous (Septum-Sealed), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Anhydrous Name Note: Septum-sealed CAS: 75-05-8 Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 Synonym: Methyl Cyanide
Acetonitrile ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: ACS Reagent Grade Molecular Formula: CH3CN Formula Weight: 41.05
Acetonitrile, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: For ACS analysis CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3CN Formula Weight: 41.05 Synonym: Methyl Cyanide
Acetonitrile Anhydrous, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Physical Form: Liquid Molecular Formula: CH3CN Linear Formula: CHCH3CN Formula Weight: 41.5 Synonym: Methyl Cyanide
Acetonitrile, Anhydrous, 99.8% min, DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Physical Form: Liquid Free From Particulates Assay Percent Range: 99.8% min. Molecular Formula: CH3CN Formula Weight: 41.05 Synonym: Methyl Cyanide
Acetonitrile, 99.5% min, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: For ACS analysis CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3CN Formula Weight: 41.05 Synonym: Methyl Cyanide
LC-MS Acetonitrile, OmniSolv, EMD Millipore
Chemical Name or Material: LC-MS Acetonitrile Name Note: LC-MS OmniSolv™ CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: CH3CN Synonym: Methyl Cyanide
Acetonitrile, DriSolv™, EMD Millipore
Chemical Name or Material: Acetonitrile Linear Formula: CH3CN Formula Weight: 41.05
  spinner