Analytical Chloroform
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™
Chemical Name or Material: Chloroform Name Note: Stabilized with Amylene CAS: 67-66-3 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, +99%, for analysis, stabilized with ethanol, ACROS Organics™
Chemical Name or Material: Chloroform CAS: 67-66-3 CAS: 64-17-5 Purity Grade: For analysis Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, Stabilized, Reagent ACS 99.8%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Stabilized CAS: 67-66-3 CAS: 64-17-5 Purity Grade: ACS Reagent Grade Assay: ≥99.8% Assay Percent Range: 99.8+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, ACS, >99.8%, Alfa Aesar™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: ACS Reagent Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 CAS: 64-17-5 Purity Grade: Extra Pure Assay: ≥99.0% (excluding stabilizer) Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, 99+%, extra pure, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Stabilized with amylene CAS: 67-66-3 CAS: 513-35-9 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 50ppm Pentene as Preservative CAS: 67-66-3 CAS: 513-35-9 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.8% Assay Percent Range: 99.8% min by GC Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform, 99.9%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Over Molecular Sieves, Stabilized CAS: 67-66-3 CAS: 513-35-9 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Water <50 ppm, Stabilized CAS: 67-66-3 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.85% Assay Percent Range: 99.9% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform Products for Science Education
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: Reagent Grade Linear Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform, 99.8%, for analysis, stabilized with amylene, ACROS Organics™
Chemical Name or Material: Chloroform Name Note: Stabilized with amylene CAS: 67-66-3 CAS: 513-35-9 Purity Grade: For analysis Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, stabilized with ethanol, for HPLC, ACROS Organics™
Chemical Name or Material: Chloroform Name Note: Stabilized with ethanol CAS: 67-66-3 CAS: 64-17-5 Purity Grade: For HPLC Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform (Approx. 0.75% Ethanol as Preservative/Technical), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 0.75% ethanol as preservative CAS: 67-66-3 CAS: 64-17-5 Purity Grade: Technical Grade Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform, Reagent ACS, Spectro Grade, 99.8%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 CAS: 64-17-5 Purity Grade: For spectroscopy Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform/isoamyl alcohol 24:1, for molecular biology, DNAse, RNAse and Protease fr, ACROS Organics™
Chemical Name or Material: Chloroform/isoamyl alcohol 24:1(v/v) Name Note: DNase, RNase and Protease Free CAS: 123-51-3 CAS: 67-66-3 CAS: 123-51-3 Purity Grade: For molecular biology Assay: Chloroform component:%gt; 99.9%, isoamyl alcohol component: 99.0% min. (by GC) MDL Number: MFCD00171265
Chloroform ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: ACS Reagent Grade Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, Macron Fine Chemicals
Chemical Name or Material: Chloroform Name Note: Purified CAS: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, J.T.Baker™
Chemical Name or Material: Chloroform CAS: 67-66-3 Physical Form: Liquid Molecular Formula: CHCl3 Formula Weight: 119.38 Synonym: Formyl trichloride
Chloroform Technical, Macron Fine Chemicals™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: Technical Grade Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Chloroform CAS: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38
ChloroForm, HPLC, J.T.Baker™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: HPLC Grade Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform ACS Reagent Grade (for General Use), Ricca Chemical
Chemical Name or Material: Chloroform CAS: 67-66-3 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Physical Form: Liquid
Chloroform Molecular Biology MP Biomedicals
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: Molecular Biology Grade Molecular Formula: CHCl3 Formula Weight: 119.38 Synonym: Formyl trichloride
ChloroForm, Reagent grade, For ACS analysis, ≥99.8%, MP Biomedicals™
Chemical Name or Material: Chloroform Name Note: For ACS analysis CAS: 67-66-3 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: ≥99.8% Molecular Formula: CHCl3 Formula Weight: 119.4 Synonym: Methylidyne trichloride
Chloroform, Anhydrous DriSolv™, EMD Millipore
Chemical Name or Material: Chloroform, Anhydrous CAS: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38 UN Number: 1888 Synonym: Trichloromethane
Chloroform, Anhydrous, 99% min., DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: DriSolv™ CAS: 67-66-3 Physical Form: Liquid Assay Percent Range: 99%, min. Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform, 99.8% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: For ACS analysis CAS: 67-66-3 Physical Form: Liquid Assay Percent Range: 99.8% min. Molecular Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform, 99% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform ACS, EMD Millipore
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: ACS Reagent Grade Linear Formula: CHCl3 Formula Weight: 119.38
  spinner