Analytical Ethanol
Ethylene glycol, Acros Organics
Chemical Name or Material: Ethylene Glycol CAS: 107-21-1 Purity Grade: Technical Grade Molecular Formula: C2H6O2 Formula Weight: 62.06 Synonym: 1, 2-Ethanediol
Ethanol, 99.5%, anhydrous, 200 proof, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: Anhydrous, 200 proof CAS: 64-17-5 CAS: 7732-18-5 Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.07 MDL Number: MFCD00003568 Synonym: Ethyl alcohol
Ethanol, 99.5%, ACS reagent, absolute, 200 proof, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: Absolute, 200 Proof CAS: 64-17-5 CAS: 7732-18-5 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.07 MDL Number: MFCD00003568 Synonym: Ethyl alcohol
Ethanol, Anhydrous (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol, Anhydrous CAS: 64-17-5 CAS: 108-10-1 CAS: 141-78-6 CAS: 67-56-1 CAS: 108-88-3 Purity Grade: Histological Grade Physical Form: Liquid MDL Number: MFCD00003568 Synonym: Ethanol, Denatured
Ethanol, 190 proof, 95%, Reagent ACS, Spectrophotometric Grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: 190 Proof CAS: 64-17-5 CAS: 7732-18-5 Purity Grade: Spectrophotometric grade Assay: ≥95.0% Assay Percent Range: 95% Molecular Formula: C2H6O Linear Formula: C2H5OH Formula Weight: 46.06 MDL Number: MFCD00003568 Synonym: Ethyl alcohol
Ethanol, 190 proof (95%), USP, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
May require a federal tax exempt certificate.
Ethanol, 200 proof (100%), Tax-Paid, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: 200 Proof (100%) CAS: 99.9 CAS Min %: 99.9 CAS Max %: 100.0 Synonym: Ethyl Alcohol
Ethanol, 140 proof (70%), Tax-Paid, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Federal Tax is included in the price
Ethanol, 190 proof (95%), Tax-Paid, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Federal Tax is included in the price, except for products 04-355-457 and 04-355-461, for which the tax is charged as a separate item. No federal permit required for purchase
Ethyl Alcohol, 200 Proof, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol, 200 Proof Name Note: 200 Proof, For ACS analysis CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3CH2OH Formula Weight: 46.07 Synonym: Ethanol
Ethyl Alcohol, 95% Denatured, 86.0 to 89.0%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 86.0 to 89.0% Molecular Formula: CH3CH2OH Formula Weight: 46.07 Synonym: Ethanol
Ethyl Alcohol, Anhydrous, 92 to 94%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol, Anhydrous CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 92 to 94% Molecular Formula: CH3CH2OH Formula Weight: 46.07 Synonym: Ethanol
Ethyl Alcohol, Anhydrous, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol, Anhydrous CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 92 to 94% Molecular Formula: CH3CH2OH Formula Weight: 46.07 Synonym: Ethanol
Ethyl Alcohol, Pure, 190 Proof, USP/NF grade, PHARMCO-AAPER
Boiling Point: 78.3°C (172.9°F) Vapor Pressure: 59.5 hPa (44.6 mmHg) at 20°C (68°F) (for 100% ethanol)
Ethyl Alcohol, ≥95%, Ultra Scientific
Chemical Name or Material: Ethyl Alcohol CAS: 000064-17-5 CAS Max %: 100.0 Assay Percent Range: >95%
Ethanol, Absolute, ENG Scientific
Chemical Name or Material: Ethanol
Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™, ACROS Organics™
Chemical Name or Material: Ethanol Name Note: Extra Dry CAS: 64-17-5 CAS: 7732-18-5 Purity Grade: Extra Dry Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.07 MDL Number: MFCD00003568
Ethyl Alcohol, 70% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ethanol CAS: Ethyl Alcohol (Ethanol) 64-17-5 CAS Min %: 60.0 CAS Max %: 72.0 CAS: Methanol (Methyl Alcohol) 67-56-1 CAS Min %: 3.0 CAS Max %: 4.0 CAS: Isopropyl Alcohol (Isopropanol, 2-Propanol) 67-63-0 CAS Min %: 3.0 CAS Max %: 4.0 CAS: Water, Deionized 7732-18-5 CAS Min %: 34.0 CAS Max %: 20.0 Physical Form: Liquid Assay Percent Range: 70% (v/v)
  spinner