Analytical Ethyl Acetate
Ethyl acetate, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Ethyl acetate Name Note: Water <50ppm CAS: 141-78-6 Purity Grade: Extra Dry Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Alfa Aesar™ Ethyl acetate, ACS, 99.5+%
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: ACS Grade Linear Formula: CH3CO2CH2CH3 Formula Weight: 88.11 MDL Number: MFCD00009171 UN Number: UN1173 Beilstein: 506104 Merck Index: 14,3757
Ethyl Acetate Products for Science Education
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: ACS Reagent Grade Linear Formula: CH3COOC2H5 Formula Weight: 88.11 Synonym: Acetic Acid Ethyl Ester
Ethyl acetate, +99.5%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate Name Note: 99.5+% CAS: 141-78-6 Purity Grade: For spectroscopy Assay: ≥99.50% Assay Percent Range: 99.5+% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl acetate, 99.9%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate Name Note: Over Molecular Sieves CAS: 141-78-6 Purity Grade: Extra Dry over Molecular Sieve Purity Grade Notes: Water <50ppm Assay: ≥99.9% (GC) Assay Percent Range: 99.9% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl Acetate (Certified ACS), Fisher Chemical
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: Certified ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic Acid Ethyl Ester
Ethyl acetate, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl Acetate 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl acetate, 99.8%, for biochemistry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: For biochemistry Assay Percent Range: 99.8% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 Synonym: Acetic acid ethyl ester
Ethyl Acetate, 99.6%, For analysis ACS, ACROS Organics™
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Ethyl Acetate, Reagent ACS, Spectro Grade, 99.5%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Purity Grade: For spectroscopy ACS Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C4H8O2 Linear Formula: CH3CO2C2H5 Formula Weight: 88.11 MDL Number: MFCD00009171 Synonym: Acetic acid ethyl ester
Alfa Aesar™ Ethyl acetate, 99%
Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Molecular Formula: C4H8O2 Linear Formula: CH3CO2CH2CH3 Formula Weight: 88.11 MDL Number: MFCD00009171 UN Number: UN1173 Beilstein: 506104 Merck Index: 14,3757 Synonym: Acetic acid ethyl ester
Ethyl Acetate, ULTRA RESI-ANALYZED™, J.T.Baker™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: ULTRA RESI-ANALYZED™ Molecular Formula: CH3COOC2H5 Formula Weight: 88.11
Ethyl Acetate UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Molecular Formula: CH3COOCH2CH3 Formula Weight: 88.11
Ethyl Acetate, HPLC Grade, J.T.Baker™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: HPLC Grade Molecular Formula: CH3COOC2H5 Formula Weight: 88.11
Ethyl Acetate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: CH3COOC2H5 Formula Weight: 88.11
Ethyl Acetate ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: ACS Reagent Grade Molecular Formula: CH3COOC2H5 Formula Weight: 88.11
Ethyl Acetate, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Name Note: Filtered through a 0.2 micron filter Purity Grade: BAKER ANALYZED™ LC-MS Reagent Molecular Formula: CH3COOC2H5 Formula Weight: 88.11
Ethyl Acetate, ≥99.5%, ACS Reagent, Honeywell™ Riedel-de Haën™
Vapor Pressure: 73mmHg (20°C) Vapor Density: 3 (20°C, vs air) Density: 0.902g/mL at 25°C Color: APHA: ≤10 Explosion Limit: 2.2 to 11.5% (38°F) Flash Point: −3°C (26.6°F) Ignition Point: 801°F Linear Formula: CH3COOC2H5 MDL Number: MFCD00009171 UN Number: UN1173 Assay: ≥99.5% Formula Weight: 88.11g/mol Purity Grade: ACS Reagent Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Beilstein: 506104 Boiling Point: 77.5°C Melting Point: −84°C
Ethyl Acetate, ACS, ≥99.5%, Honeywell™ Riedel-de Haën™
Assay: ≥99.5% Formula Weight: 88.11g/mol Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Beilstein: 506104 MDL Number: MFCD00009171 UN Number: UN1173
Ethyl Acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Vapor Pressure: 73mmHg (20°C) Vapor Density: 3 (20°C, vs air) Density: 0.902g/mL at 25°C Color: APHA: >10 Explosion Limit: 2.2 to 11.5% (38°F) Flash Point: −3°C (26.6°F) Ignition Point: 801°F Physical State: Liquid Color: APHA: >10 Odor: Like fruit Boiling Point: 77°C Melting Point: −84°C Solubility: ∽90g/L
Ethyl Acetate, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Assay: ≥99.5% (GC) Formula Weight: 88.11g/mol Chemical Name or Material: Ethyl acetate CAS: 141-78-6 Beilstein: 506104 MDL Number: MFCD00009171 UN Number: UN1173
Ethyl Acetate, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate Name Note: For ACS analysis CAS: 141-78-6 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3COOCH2CH3 Formula Weight: 88.11 Synonym: Acetic ester
Anhydrous Ethyl Acetate, 99.5% (ACS Guaranteed Reagent), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate, 99.5% CAS: 141-78-6 Purity Grade: ACS Reagent Grade Physical Form: Anhydrous Formula Weight: 88.11
Ethyl Acetate, DriSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate Name Note: DriSolv™ Purity Grade: DriSolv Physical Form: Liquid Linear Formula: CH3COOCH2CH3 Formula Weight: 88.11
Ethyl Acetate ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Acetate CAS: 141-78-6 Purity Grade: ACS Reagent Grade Linear Formula: CH3COOCH2CH3 Formula Weight: 88.11
Ethyl Acetate, EMPLURA™, EMD Millipore™
CAS: 141-78-6 Molecular Formula: CH3COOC2H5 Formula Weight: 88.11 Chemical Name or Material: Ethyl Acetate
  spinner