Analytical IPA Isopropanol
2-Propanol (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol
IPA (Isopropyl Alcohol), 70% v/v, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: IPA (Isopropyl Alcohol), 70% v/v CAS: 7732-18-5 CAS: 67-63-0 Assay: 68.0 to 72.0% Assay Percent Range: Assay: 68.0 to 72.0% Molecular Formula: C3H8O Formula Weight: 60.1 MDL Number: MFCD00011674
Isopropanol, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: Reag. Ph. Eur. CAS: 67-63-0 Purity Grade: For analysis Assay: ≥99.0% Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: Extra Dry Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: Over Molecular Sieves CAS: 67-63-0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.5+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: 99.8+% CAS: 67-63-0 Purity Grade: Pure Assay: ≥99.50% (GC) Assay Percent Range: 99.5+% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
2-Propanol (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Laboratory Grade Purity Grade Notes: Purified Physical Form: Liquid Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol
Isopropyl Alcohol Products for Science Education
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Assay Percent Range: >99% Linear Formula: (CH3)2CHOH Formula Weight: 6.1 Synonym: Isopropanol
Isopropanol, 99.5+%, extra pure, ACROS Organics™
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: Extra Pure Assay: ≥99.50% Assay Percent Range: 99.5+% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, specified according to the requirements of Ph. Eur., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol, specified according to the requirements of Ph. Eur. Assay: ≥99.0% (GC) Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.5%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: For spectroscopy ACS Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
2-Propanol (Low Water), Fisher Chemical
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Low Water Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H8O Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol, Isopropyl Alcohol
Isopropanol, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: Extra Dry Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 70% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Isopropanol CAS: 7732-18-5 CAS Min %: 29.0 CAS Max %: 31.0 CAS: 67-63-0 CAS Min %: 69.0 CAS Max %: 71.0 Synonym: 2-Propanol
2-Propanol, HPLC, J.T.Baker™
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: HPLC Grade Molecular Formula: CH3CHOHCH3 Formula Weight: 60.1 Synonym: IPA
Isopropyl Alcohol, AR, For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Isopropyl Alcohol Name Note: ACS CAS: 67-63-0 Purity Grade: ACS Reagent Grade Physical Form: Liquid Assay Percent Range: 99.5 % min. Molecular Formula: CH3CHOHCH3 Formula Weight: 60.1
Isopropyl Alcohol, ACS, For Histological Use, Macron Fine Chemicals™
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Purity Grade: ACS Reagent Grade Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: 2-Propanol
Isopropanol, 30% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Isopropanol CAS: 7732-18-5 CAS Min %: 69.0 CAS Max %: 71.0 CAS: 67-63-0 CAS Min %: 29.0 CAS Max %: 31.0 Physical Form: Liquid
Isopropanol, 90% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Isopropanol CAS: 7732-18-5 CAS Min %: 9.0 CAS Max %: 11.0 CAS: 67-63-0 CAS Min %: 89.0 CAS Max %: 91.0 Physical Form: Liquid
Isopropanol, 10% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Isopropanol CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 91.0 CAS: 67-63-0 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Liquid
Isopropanol ACS Reagent Grade (for General Use), Ricca Chemical
Chemical Name or Material: Isopropanol CAS: 67-63-0 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Physical Form: Liquid
Isopropanol, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Isopropanol Name Note: For Laboratory Cleaning and Disinfecting CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 67-63-0 CAS Min %: 49.0 CAS Max %: 51.0
Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals
Chemical Name or Material: Isopropyl Alcohol, 99% CAS: 67-63-0 Formula Weight: 60.09
Isopropyl alcohol, ≥99%, Reagent grade, For ACS analysis, MP Biomedicals™
Chemical Name or Material: Isopropyl alcohol Name Note: For ACS analysis CAS: 67-63-0 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: ≥99% Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: 2-propanol
Isopropyl Alcohol, 99% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Molecular Formula: CH3CHOHCH3 Linear Formula: CH3CHOHCH3 Formula Weight: 6.1 Synonym: 2-Propanol
Isopropyl Alcohol, 70%, Reagent, EMD Millipore
Chemical Name or Material: Isopropyl Alcohol, 70% CAS: 67-63-0 Purity Grade: Reagent Grade
iso-Propyl Alcohol ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: iso-Propyl Alcohol Purity Grade: ACS Reagent Grade Linear Formula: CH3CHOHCH3 Formula Weight: 60.1
Isopropyl Alcohol, GR, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Physical Form: Liquid Molecular Formula: C3H8O Formula Weight: 60.11 Synonym: 2-Propanol
Isopropyl Alcohol, 70%, Reagent grade, EMD Millipore
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Synonym: 2-Propanol
  spinner