Analytical Methanol
Methanol, 99.8+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: ACS Reagent Grade Assay: ≥99.80% Assay Percent Range: 99.8+% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.9%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol Products for Science Education
Chemical Name or Material: Methanol CAS: 67-56-1 Linear Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl Alcohol
Methanol, 99.9%, for analysis, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: For analysis Assay: ≥99.8% Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.9%, for spectroscopy, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: For spectroscopy Assay: ≥99.85% Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: water <50 ppm CAS: 67-56-1 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.8% (GC) Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, ACS, absolute, low acetone, >99.8%, Alfa Aesar™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™
Chemical Name or Material: Methanol, Anhydrous CAS: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Methanol Name Note: Anhydrous CAS: 67-56-1 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C H4 O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Laboratory Grade Physical Form: Liquid Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Technical Grade Molecular Formula: CH4O Formula Weight: 32.04 Synonym: Methyl alcohol
Methanol (Low Water), Fisher Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Low Water Purity Grade Notes: Also meets ACS specifications Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, Anhydrous, 99%, Alfa Aesar
Chemical Name or Material: Methanol CAS: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04
Methanol, 99.8%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: ACS Grade Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 90% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Methanol CAS: 7732-18-5 CAS Min %: 9.0 CAS Max %: 11.0 CAS: 67-56-1 CAS Min %: 89.0 CAS Max %: 91.0 Physical Form: Liquid
Methanol/Toluene Solution, 1:1 (50% Methyl Alcohol, 50% Toluene), Ricca Chemical
Chemical Name or Material: Methanol/Toluene Solution CAS: 108-88-3 CAS Min %: 51.0 CAS Max %: 53.0 CAS: 67-56-1 CAS Min %: 46.65 CAS Max %: 48.55 Physical Form: Liquid
Methanol, ACS Reagent Grade, Absolute, Ricca Chemical
Chemical Name or Material: Methanol Name Note: Absolute CAS: 67-56-1 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Physical Form: Liquid
Methanol, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Methanol CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 67-56-1 CAS Min %: 49.0 CAS Max %: 51.0 Physical Form: Liquid Synonym: Methyl Alcohol
Methyl Alcohol ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Alcohol Name Note: Anhydrous CAS: 67-56-1 Purity Grade: ACS Reagent Grade Molecular Formula: CH3OH Formula Weight: 32.04
Methyl Alcohol , Macron Fine Chemicals™
Chemical Name or Material: Methyl Alcohol Name Note: Anhydrous CAS: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04
Methanol ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: ACS/NF/USP Linear Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl Alcohol
Methanol, Anhydrous, DriSolv™, Septum-Sealed, EMD Millipore
Chemical Name or Material: Methanol Name Note: Septum-sealed CAS: 67-56-1 Physical Form: Liquid Free From Particulates Assay Percent Range: 99.8% min. Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl Alcohol
Methanol, 99.8% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: For ACS analysis CAS: 67-56-1 Physical Form: Liquid Assay Percent Range: 99.8% min. Molecular Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl Alcohol
Methanol Anhydrous ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: Anhydrous Purity Grade: ACS Reagent Grade Linear Formula: CH3OH Formula Weight: 32.04 Water: 0.03% max
Methanol, anhydrous, ACS, GR, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol, anhydrous Name Note: For ACS analysis CAS: 67-56-1 Physical Form: Liquid Molecular Formula: CH4 Formula Weight: 32 Synonym: Methyl Alcohol
Methanol, OmniSolv™, EMD Millipore
Chemical Name or Material: Methanol CAS: 67-56-1 Molecular Formula: CH4O Formula Weight: 32.04 UN Number: 1230 Synonym: Methyl Alcohol
  spinner