Analytical Methylene Chloride
Dichloromethane, 99.5%, for spectroscopy ACS, stabilized with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: 99.5% CAS: 75-09-2 CAS: 513-35-9 Purity Grade: ACS Grade Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottle, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Purity Grade Notes: Water <30ppm Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane 99+%, stabilized with amylene, pure, ACROS Organics™
Chemical Name or Material: Dichloromethane Assay Percent Range: 99+% Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Purity Grade Notes: Water <30ppm Assay: ≥99.85% (GC) Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.8%, for spectroscopy, stabilized with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Stabilized CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For spectroscopy Assay: ≥99.75% Assay Percent Range: 99.8% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.6%, ACS reagent, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Stabilized with Amylene CAS: 75-09-2 CAS: 513-35-9 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For analysis Assay: ≥99.80% Assay Percent Range: 99.8+% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Anhydrous, SC CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene Chloride
Dichloromethane, 99.9%, Extra Dry, AcroSeal™, 99.9%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Anhydrous CAS: 75-09-2 CAS: 513-35-9 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.9% Molecular Formula: C H2 Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride, PHOTREX™ Reagent, J.T.Baker™
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: PHOTREX™ Reagent Molecular Formula: CH2Cl2 Formula Weight: 84.93
Methylene Chloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: Baker Analyzed™ ACS Reagent Purity Grade Notes: Meets ACS reagent chemical requirements Molecular Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Purity Grade: ACS Reagent Grade Molecular Formula: CH2Cl2 Formula Weight: 84.93
Methylene Chloride, HPLC Grade, J.T.Baker™
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: HPLC Grade Molecular Formula: CH2Cl2 Formula Weight: 84.93
Methylene Chloride, Low water, BakerDRY™, J.T.Baker™
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Name Note: Low water Purity Grade: BakerDRY™ Molecular Formula: CH2Cl2 Formula Weight: 84.93
Methylene Chloride (Pentene Preservative), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Pentene Preservative CAS: 110-83-8 CAS: 513-35-9 CAS: 67-56-1 CAS: 75-09-2 Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: DCM
Methylene chloride, ≥99.8%, Ultra Resi-analyzed, J.T.Baker™
Chemical Name or Material: Methylene chloride CAS: 75-09-2 Purity Grade: ULTRA RESI-ANALYZED™ Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Dichloromethane
Dichloromethane, puriss. p.a., ACS Reagent, Reag. ISO, ≥99.9% (GC), Honeywell™ Riedel-de Haën™
Appearance: Liquid Color: Colorless Vapor Pressure: 6.83psi (20°C), 24.45psi (55°C) Vapor Density: 2.9 (vs air) Density: 1.325g/mL at 25°C, 1.320 to 1.330g/Lat 20°C Explosion Limit: 0.22 Ignition Point: 1223°F Residue after Evaporation: 0 to 0.001% Melting Point: −97°C Boiling Point: 39.8° to 40°C Refractive Index: n20/D 1.4235 to 1.4245 Titratable Acid: Free acid: ≤0.001% (as HCl) Stabilizer: 10 to 60ppm amylene
Methylene Chloride, ULTRA RESI-ANALYZED, J.T.Baker™
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: ULTRA RESI-ANALYZED™ Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: dichloromethane
Dichloromethane, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: For ACS analysis CAS: 75-09-2 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene Chloride
Dichloromethane, ACS, GR, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: For ACS analysis CAS: 75-09-2 Physical Form: Liquid Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene Chloride
Dichloromethane (HR-GC), OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ™OmniSolv™ Dichloromethane Name Note: HR-GC, OmniSolv™ CAS: 75-09-2 Purity Grade: HR-GC Physical Form: Liquid Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene Chloride
Methylene Chloride, ≥95%, Ultra Scientific
Chemical Name or Material: Methylene Chloride CAS: 000075-09-2 Physical Form: Liquid Assay Percent Range: >95%
  spinner