Analytical Toluene
Toluene, 99.5%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, 99.5%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: 99.5% CAS: 108-88-3 Purity Grade: ACS Grade Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C7H8 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
Chemical Name or Material: Toluene Name Note: Anhydrous CAS: 108-88-3 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C7 H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, ACS, 99.5%, Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 99.5% Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Laboratory Grade Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, anhydrous, 99.8%, Alfa Aesar™
Chemical Name or Material: Toluene, anhydrous CAS: 108-88-3 Physical Form: Liquid Assay Percent Range: 99.8% Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Alfa Aesar™ Toluene, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™
Chemical Name or Material: Toluene, anhydrous CAS: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: water <50 ppm CAS: 108-88-3 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.8% (GC) Assay Percent Range: 99.85% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene Products for Science Education
Chemical Name or Material: Toluene CAS: 108-88-3 Linear Formula: CH3C6H5 Formula Weight: 92.14 Synonym: Tol, Methylbenzene
Toluene, 99.8+%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: For analysis Assay: ≥99.80% Assay Percent Range: 99.8+% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, 99.85%, Extra Dry, AcroSeal™, package of 4x25ML bottles, ACROS Organics™
Chemical Name or Material: Toluene Name Note: Extra Dry CAS: 108-88-3 Purity Grade: Extra Dry Assay: ≥99.8% (GC) Assay Percent Range: 99.85% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 Synonym: Methylbenzene
Toluene, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: Water < 50ppm CAS: 108-88-3 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C7 H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Toluene CAS: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14
Toluene, Purified, Macron Fine Chemicals
Chemical Name or Material: Toluene Name Note: Purified CAS: 108-88-3 Physical Form: Liquid Molecular Formula: C7H8 Formula Weight: 92.14
Toluene ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: ACS Reagent Grade Molecular Formula: C6H5CH3 Formula Weight: 92.14
Toluene ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: ACS Reagent Grade Linear Formula: C6H5CH3 Formula Weight: 92.14
Toluene, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: For ACS analysis CAS: 108-88-3 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: C6H5CH3 Formula Weight: 92.14 Synonym: Methylbenzene
Toluene, Dried, SeccoSolv™, EMD Millipore™
CAS: 108-88-3 Melting Point: −95°C Boiling Point: 110.6°C (1013 hPa)
Toluene, Anhydrous, 99.8% min., DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene, Anhydrous Name Note: DriSolv™ CAS: 108-88-3 Physical Form: Liquid Assay Percent Range: 99.8%, min. Molecular Formula: C6H5CH3 Linear Formula: C6H5CH3 Formula Weight: 92.14 Synonym: Methylbenzene
Toluene, GMP/ACS GRADE, PHARMCO-AAPER
Chemical Name or Material: Toluene CAS: 108-88-3 Physical Form: Liquid Molecular Formula: C7H8 Formula Weight: 92.14 Synonym: Methylbenzene
  spinner