Analytical Water -Plasma Grade
Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 7732-18-5 Certificate of Analysis: Certificate of Lot Analysis included with each bottle. ChemAlert Storage Symbol: Gray Chemical Name or Material: Water Density: 1.000g/cm³ Boiling Point: 100°C Color: Clear Colorless Formula Weight: 18.02 Melting Point: 0°C pH: 7 Physical Form: Liquid Purity Grade: Ultra Trace Elemental Analysis Grade MDL Number: 11332 Molecular Formula: H2O Packaging: LDPE Bottle Vapor Pressure: 17.5mmHg at 20°C
Water ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Water CAS: 7732-18-5 Molecular Formula: H2O Formula Weight: 18.02
  spinner