Application Specific Acetone
Acetone, for residue analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: ECD Tested for Pesticide Analysis CAS: 67-64-1 Purity Grade: For residue analysis Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.9%, for residue analysis, for trace analysis of polyaromatic hydrocarbon, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: Tested for Trace Analysis of Polyaromatic Hydrocarbons CAS: 67-64-1 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, Spectrophotometric Grade, 99.5+%, Alfa Aesar
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: C3H6O Formula Weight: 58.08 MDL Number: MFCD00008765
Acetone (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.5% Assay Percent Range: 99.5% min by GC Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: 99+% CAS: 67-64-1 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3 H6 O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (Electronic), Fisher Chemical
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Electronic Grade Assay: ≥99.5% Assay Percent Range: 96% min by GC Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, Environmental Grade, >99.5% Alfa Aesar™
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: C3H6O Formula Weight: 58.08 MDL Number: MFCD00008765
Acetone, Semiconductor Grade, 99.5%, Alfa Aesar
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Semiconductor Grade Physical Form: Liquid Assay Percent Range: 99.5% Molecular Formula: C3H6O Formula Weight: 58.08 MDL Number: MFCD00008765
Acetone, HPLC Grade, 99.5+%, Alfa Aesar
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: C3H6O Formula Weight: 58.08 MDL Number: MFCD00008765
Acetone, 99.8%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.9%, for residue analysis, ECD tested halocarbons free grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: ECD Tested CAS: 67-64-1 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: 99.9 to 100% (on anhydrous basis) Assay Percent Range: 99.9% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, 99.5%, for electronic use, residue free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: Residue Free CAS: 67-64-1 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For electronic use Assay: ≥99.8% Assay Percent Range: 99.5% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.5% Assay Percent Range: 99.9% min by GC Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (Pesticide), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Pesticide Grade Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: Histological Grade Physical Form: Liquid Assay Percent Range: Assay: ≥99.5% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (GC Resolv™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: GC Resolv Assay: ≥99.5% Assay Percent Range: ≥99.5% Molecular Formula: C3H6O Linear Formula: CH3COCH3 Formula Weight: 58.08 MDL Number: MFCD00008765 Synonym: 2-Propanone
Acetone (UltimAR), Macron Fine Chemicals™
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: UltimAR Molecular Formula: CH3COCH3 Formula Weight: 58.08
Acetone (ChromAR), Macron Fine Chemicals™
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: ChromAR™ Molecular Formula: CH3COCH3 Formula Weight: 58.08
Acetone (OmniSolv™) 99.5%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone, 99.5% CAS: 67-64-1 Purity Grade: OmniSolv™ Assay: ≥99.5% (GC, corrected for water content) Linear Formula: CH3COCH3 Formula Weight: 58.08 Synonym: Dimethyl Ketone
Acetone, Histology grade, 99% min, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone Name Note: For analysis Histology CAS: 67-64-1 Physical Form: Liquid Assay: ≥99% (GC) Assay Percent Range: 99% min. Molecular Formula: CH3COCH3 Formula Weight: 58.08 Synonym: Dimethyl Ketone
Acetone, HR-GC, 99.7% min, OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: HR-GC Acetone Name Note: HR-GC, OmniSolv™ CAS: 67-64-1 Physical Form: Liquid Assay Percent Range: 99.7%, min. Molecular Formula: CH3COCH3 Linear Formula: CH3COCH3 Formula Weight: 58.08 Synonym: Dimethyl Ketone
Acetone, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetone CAS: 67-64-1 Purity Grade: HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs Assay: ≥99.5% (GC, corrected for water content) Linear Formula: CH3COCH3 Formula Weight: 58.08
Acetone, for gas chromatography ECD and FID SupraSolv™, EMD Millipore
Chemical Name or Material: Acetone CAS: 67-64-1 Molecular Formula: CH3COCH3 Formula Weight: 58.08 UN Number: 1090 Synonym: Dimethyl ketone, Propanone, 2-Propanone
Acetone, for gas chromatography MS SupraSolv™, EMD Millipore
Chemical Name or Material: Acetone CAS: 67-64-1 Molecular Formula: CH3COCH3 Formula Weight: 58.08 UN Number: 1090 Synonym: Propanone
Acetone, for Spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: Acetone CAS: 67-64-1 Molecular Formula: CH3COCH3 Formula Weight: 58.08 UN Number: 1090 Synonym: Dimethyl ketone, Propanone, 2-Propanone
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Beilstein: 635680 CAS: 67-64-1 Chemical Name or Material: Acetone Density: 0.791g/mL�at 25°C Assay: ≥99.8% Boiling Point: 56°C/760mmHg Formula Weight: 58.08g/mol Melting Point: −94°C Purity Grade: CHROMASOLV™ EINECS Number: 200-662-2 Health Hazard 2: P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235 Health Hazard 3: H225-H319-H336 Linear Formula: CH3COCH3 MDL Number: MFCD00008765 Molecular Formula: C3H6O Name Note: for HPLC Refractive Index: n20/D 1.359 RTECS Number: AL3150000 Vapor Density: 2 (vs. air) Vapor Pressure: 184mmHg (20°C)
  spinner