Application Specific Acetonitrile
Alfa Aesar™ Acetonitrile, LC-MS Grade, 99.8%
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: LC/MS Grade Formula Weight: 41.05 MDL Number: MFCD00001878 UN Number: UN1648 Beilstein: 741857 Merck Index: 14,70 Synonym: Methyl cyanide
Acetonitrile (HPLC), Fisher Bioreagents™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: ACS/HPLC Purity Grade Notes: Also meets ACS specifications Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 Synonym: Ethanenitrile
Acetonitrile (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: Assay: 99.9% min by GC Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: ACN
Acetonitrile, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.9%, for residue analysis, ECD tested, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: ECD tested, For pesticide analysis CAS: 75-05-8 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, HPLC Grade (Far UV), >99.8%, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC (Far UV) Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile (Pesticide), Fisher Chemical
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Pesticide Grade Purity Grade Notes: Also meets ACS specifications Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Ethanenitrile
Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 75-05-8 Color: Colorless Physical Form: Liquid Molecular Formula: C2H3N
Acetonitrile, Optima UHPLCMS, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: UHPLC-MS Optima Physical Form: Liquid Assay Percent Range: 99.9% minimum by GC Molecular Formula: C2H3N
Acetonitrile, Spectrophotometric Grade, 99.7%, Alfa Aesar™
Chemical Name or Material: Acetonitrile Name Note: Meets ACS spectrophotometric requirements. Filtered through 0.2μfilters CAS: 75-05-8 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, 99.9%, for residue analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: Tested for Trace Analysis of polyaromatic Hydrocarbons CAS: 75-05-8 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 CAS Min %: 99.85 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, 99.8%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: For HPLC Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, HPLC Grade, 99.7+%, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC Grade Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 UN Number: UN1648 Beilstein: 741857 Merck Index: 14,70 Synonym: Methyl cyanide
Acetonitrile, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.9% Assay Percent Range: Assay: 99.9% min by GC Molecular Formula: C2H3N Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: ACN
Acetonitrile, HPLC Grade, 99.7+% min, Alfa Aesar™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% min. Molecular Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Methyl cyanide
Acetonitrile, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile Name Note: 99.9% CAS: 75-05-8 Purity Grade: For HPLC Assay: ≥99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile (Certified), Fisher Chemical
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: Certified Grade Assay: ≥99.9% Assay Percent Range: ≥98.5 % Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.05 MDL Number: MFCD00001878 Synonym: Ethanenitrile
Acetonitrile, for biochemistry, synthesis grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: For biochemistry Assay: ≥99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
Acetonitrile, LC/MS grade, ACROS Organics™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: LC/MS Grade Assay: ≥99.9% Molecular Formula: C2H3N Linear Formula: CH3CN Formula Weight: 41.04 MDL Number: MFCD00001878 Synonym: AN
80% Acetonitrile, 20% Water with 0.1% Formic Acid, Optima LC/MS, Fisher Chemical
Chemical Name or Material: Acetonitrile with formic acid CAS: 75-05-8 CAS: 7732-18-5 CAS: 64-18-6 Purity Grade: Optima LC/MS
Acetonitrile, BAKER ANALYZED HPLC ULTRA Gradient Solvent, J.T.Baker™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05
Acetonitrile UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05
Acetonitrile ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05
Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Purity Grade: DNA Synthesis Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C2H3N Formula Weight: 41.05
Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™
CAS: 75-05-8 Chemical Name or Material: Acetonitrile Density: 0.786g/mL at 25°C Assay: ≥99.9% Color: Colorless Formula Weight: 41.05 Explosion Limit: 0.16 Flash Point: 2°C (35.6°F) Ignition Point: 973°F Linear Formula: CH3CN MDL Number: MFCD00001878 UN Number: UN1648 Vapor Density: 1.41 (vs air) Vapor Pressure: 72.8 mmHg (20°C)
Acetonitrile, OmniSolv™, Non−UV, EMD Millipore™
CAS: 75-05-8 Molecular Formula: CH3CN Formula Weight: 41.05 Chemical Name or Material: Acetonitrile
Acetonitrile, Gradient grade, 99.9% min, OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Acetonitrile CAS: 75-05-8 Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: CH3CN Formula Weight: 41.05 Synonym: Methyl Cyanide
  spinner