Application Specific Ethanol
Alfa Aesar™ Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK
Chemical Name or Material: Ethanol CAS: 64-17-5 Name Note: Anhydrous, denatured Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.08 MDL Number: MFCD00003568 UN Number: UN1993 Beilstein: 1718733 Merck Index: 14,3760 Synonym: Ethyl alcohol
Ethanol Solution 96%, Molecular Biology Grade, Regulated, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 96% Ethanol Solution Purity Grade: Molecular Biology Grade Synonym: 96% Ethyl Alcohol Solution CAS: 64-17-5 (Ethanol) CAS: 7732-18-5 (Water)
Ethanol, denatured, Spectrophotometric Grade, 90%, Alfa Aesar™
Chemical Name or Material: Ethanol, denatured CAS: 64-17-5 CAS: 67-56-1 CAS: 67-63-0 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 90% Molecular Formula: C2H6O Formula Weight: 46.08 MDL Number: MFCD00003568
Alfa Aesar™ Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v)
Chemical Name or Material: Ethanol CAS: 64-17-5 Name Note: Denatured Formula Weight: 46.08 MDL Number: MFCD00003568 UN Number: UN1170 Beilstein: 1718733 Merck Index: 14,3760 Synonym: Ethyl alcohol
Ethanol, 200 proof, Molecular Biology Grade, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Pure Ethanol products are taxed according to the government schedule of excise taxes detailed in US CFR Part 27 (211.1119). This excise tax is included in the cost of this product. There is no need for Federal Permits
Ethanol, 70% Solution, Molecular Biology Grade, Denatured, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 70% Denatured Ethanol Solution (Molecular Biology Grade) CAS: 64-17-5 CAS: 67-63-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Synonym: 70% Denatured Ethyl Alcohol Solution
Ethanol Solution 70%, Molecular Biology Grade, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Physical Form: Liquid Chemical Name or Material: 70% Ethanol Solution Purity Grade: Molecular Biology Grade Ethanol: 68%-72% (GC-TCD) Water: 28%-32% (GC-TCD) Color: Clear Colorless RNase: Not Detected DNase: Not Detected Protease: Not Detected
Reagent Alcohol, 13.5% (v/v) Denatured Alcohol, Ricca Chemical
Chemical Name or Material: Reagent Alcohol Name Note: Denatured Alcohol Assay Percent Range: 13.5% (v/v)
Reagent Alcohol, 18.5% (v/v) Denatured Alcohol, Ricca Chemical
Chemical Name or Material: Reagent Alcohol Name Note: Denatured Alcohol Assay Percent Range: 18.5% (v/v)
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: Absolute (200 Proof) CAS: 64-17-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: Assay: ≥99.5% (v/v) Molecular Formula: C2H6O Linear Formula: C2H5OH Formula Weight: 46.07 Synonym: Ethyl Alcohol
Ethanol for Spectroscopy Uvasol™ EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol CAS: 64-17-5 Molecular Formula: C2H6O Formula Weight: 46.07 UN Number: 1170 Synonym: EtOH
Ethyl Alcohol Non UV, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol Name Note: Non UV, Denatured CAS: 64-17-5 Physical Form: Liquid Linear Formula: CH3CH2OH + denaturants Formula Weight: 46.7 Synonym: Ethanol
Ethyl Alcohol, Pure, 200 Proof, OmniPur™, EMD Millipore
Chemical Name or Material: Ethyl Alcohol Molecular Formula: C2H6O Formula Weight: 46.08 UN Number: 1170 Synonym: Ethanol, Alcohol
  spinner