Application Specific IPA Isopropanol
Alfa Aesar™ 2-Propanol, Spectrophotometric Grade, 99.7+%
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Spectrophotometric grade Formula Weight: 60.1 MDL Number: MFCD00011674 UN Number: UN1219 Beilstein: 635639 Merck Index: 14,5208
Isopropanol, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.8%, for electronic use, residue free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: Residue Free CAS: 67-63-0 Purity Grade: For electronic use Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, 99.5%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: DNAse, RNAse and Protease Free CAS: 67-63-0 Purity Grade: For molecular biology Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
2-Propanol, Alfa Aesar™
Chemical Name or Material: 2-Propanol Name Note: GC Grade CAS: 67-63-0 Purity Grade: GC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674
Isopropanol, 99%, for biochemistry and histology, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: For biochemistry and histology Assay Percent Range: 99% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 Synonym: 2-Propanol
Isopropanol 99.5%, for HPLC, ACROS Organics™
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: HPLC Grade Assay Percent Range: 99.5% Molecular Formula: C3H8O Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
2-Propanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Certified Grade Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol
Isopropanol (IPA), Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent Assay: ≥99.9% Assay Percent Range: Assay: 99.9% min by GC Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: IPA
Isopropanol, 99.9%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol Name Note: 99.9% CAS: 67-63-0 Purity Grade: For HPLC Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
2-Propanol (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Histological Grade Physical Form: Liquid Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: IPA
2-Propanol (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: C3H8O Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: IPA
2-Propanol (Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H8O Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol
Isopropanol, 99.5%, for HPLC gradient grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: For HPLC gradient grade Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: 2-Propanol
Isopropanol, Optima™LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Additional Information: Vapor Pressure: 43mmHg at 20°C Aluminum (Al): 10 ppb max. Assay Percent Range: Assay: ≥99.90% Calcium (Ca): 10 ppb max. CAS: 67-63-0 Certificate of Analysis: Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request Chemical Name or Material: 2-Propanol Copper (Cu): 5 ppb max. Density: 2.1 (Air = 1.0) Assay: ≥99.90% Boiling Point: 81°C Color: Colorless Formula Weight: 60.1 Melting Point: −89.5°C pH: 7 Physical Form: Liquid Purity Grade: Optima™ LC/MS Identification: Pass Test Iron (Fe): 5 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 5 ppb max. Linear Formula: (CH3)2CHOH Magnesium (Mg): 5 ppb max. Manganese (Mn): 5 ppb max. Merck Index: 15, 5254 MDL Number: 11674 Molecular Formula: C3H8O Nickel (Ni): 5 ppb max. Packaging: Glass Bottle Physical State: Liquid Potassium (K): 10 ppb max. Purity Grade Notes: Also meets ACS specifications Residue after Evaporation: 1ppm max. Silver (Ag): 5 ppb max. Sodium (Na): 50 ppb max. Titratable Acid or Base: 0.0001 mEq Acid or Base max. Vapor Density: 2.1 at 20 °C / 68 °F Vapor Pressure: 43mmHg at 20°C Viscosity: 2.27 mPaS at 20°C Water: 0.05% max. Zinc (Zn): 10 ppb max.
2-Propanol (Pesticide), Fisher Chemical
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: Pesticide Grade Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol
Isopropyl Alcohol ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Molecular Formula: CH3CHOHCH3 Formula Weight: 60.1
Isopropanol, Molecular Biology Grade, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol CAS: 67-63-0 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 Synonym: 2-Propanol
Isopropanol, 99.9% (HPLC Grade), Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Isopropanol, 99.9% CAS: 67-63-0 Purity Grade: HPLC Grade Purity Grade Notes: Also meets ACS specifications Assay: ≥99.9% Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 Synonym: 2-Propanol
Isopropanol, 70%, Honeywell™
Chemical Name or Material: Isopropanol CAS: 67-63-0 MDL Number: MFCD00011674 UN Number: UN1219
2-Propanol, Laboratory Reagent, ≥99.5%, Honeywell™
Solubility: Soluble in water Vapor Pressure: 33mmHg (20°C), 44mmHg (25°C) Vapor Density: 2.1 (vs air) Density: 0.785g/mL at 25°C Explosion Limit: 0.02 Flash Point: 12°C (53.6°F) Ignition Point: 750°F Linear Formula: (CH3)2CHOH MDL Number: MFCD00011674 UN Number: UN1219 Assay: ≥99.5% Formula Weight: 60.1g/mol Purity Grade: Laboratory Reagent Chemical Name or Material: 2-Propanol CAS: 67-63-0 Beilstein: 635639 Boiling Point: 77° to 83°C at 1013 hPa Melting Point: -90°C Refractive Index: n20/D 1.377(lit.) Physical State: Liquid Color: Colorless
2-Propanol, ≥99.8% (GC), Honeywell Riedel-de Haën™
CAS: 67-63-0 Chemical Name or Material: 2-Propanol Density: 0.785g/mL at 25°C Assay: ≥99.8% (GC) Color: Colorless Formula Weight: 60.1 Purity Grade: CHROMASOLV™ Explosion Limit: 0.02 Flash Point: 12°C (53.6°F) Ignition Point: 750°F Linear Formula: (CH3)2CHOH MDL Number: MFCD00011674 UN Number: UN1219 Vapor Density: 2.1 (vs air) Vapor Pressure: 33 mmHg (20°C), 44 mmHg (25°C)
iso-Propyl Alcohol, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: iso-Propyl Alcohol CAS: 67-63-0 Purity Grade: HPLC Grade Linear Formula: CH3CHOHCH3 Formula Weight: 60.1 Synonym: 2-Propanol
2-Propanol For gas chromatography ECD and FID SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Molecular Formula: C3H8O Formula Weight: 60.1 UN Number: 1219 Synonym: Isopropyl alcohol, Isopropanol, IPA
2-Propanol For spectroscopy Uvasol™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Molecular Formula: C3H8O Formula Weight: 60.1 UN Number: 1219 Synonym: Isopropyl alcohol, Isopropanol, IPA
Isopropyl Alcohol, OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: OmniSolv ™Isopropyl Alcohol Name Note: OmniPure™ CAS: 67-63-0 Linear Formula: CH3CHOHCH3 Synonym: 2-Propanol
  spinner