Application Specific Methanol
Methanol, LC/MS grade, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Assay: ≥99.9% Molecular Formula: C H4 O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, Optima™ LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Additional Information: Vapor Pressure: 128hPa at 20°C Aluminum (Al): 10 ppb max. Assay Percent Notes: GC Assay Percent Range: Assay: 99.9% min by GC Barium (Ba): 10 ppb max. Cadmium (Cd): 10 ppb max. Calcium (Ca): 20 ppb max. CAS: 67-56-1 Certificate of Analysis: Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request Chemical Name or Material: Methanol Chromium (Cr): 10 ppb max. Cobalt (Co): 10 ppb max. Copper (Cu): 10 ppb max. Density: 0.791g/cm³ Assay: ≥99.9% Boiling Point: 64.7°C Color: Colorless Formula Weight: 32.04 Melting Point: −98°C Physical Form: Liquid Purity Grade: Optima™ LC/MS Filtered Through: 0.1-micron filtered. Identification: Pass Test Impurities: ≤10ppb (Ionic, Potassium) Iron (Fe): 10 ppb max. LCMS at Negative Mode as Aldicarb: 50 ppb max. LCMS at Positive Mode as Reserpine: 50 ppb max. LCMS Gradient Suitability: ≤2 mAU - PDA (200-400nm) LCMS Negative Mode as Mecoprop: ≤300ppb LCMS Positive Mode as Propazine: ≤100ppb Lead (Pb): 10 ppb max. Linear Formula: CH3OH Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 6029 MDL Number: 4595 Molecular Formula: CH4O Nickel (Ni): 10 ppb max. Physical State: Liquid Potassium (K): 10 ppb max. Purity Grade Notes: Formulated for UHPLC-UV Residue after Evaporation: ≤1ppm Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Tin (Sn): 10 ppb max. Titratable Acid: ≤0.0003meq/g Titratable Base: ≤0.0002meq/g Vapor Density: 1.11 Vapor Pressure: 128 hPa at 20°C Viscosity: 0.55cp at 20°C Water: ≤0.02% Zinc (Zn): 10 ppb max.
Thiazole-5-methanol, 95%, Acros Organics™
Density: 1.3200g/mL Formula Weight: 115.15 CAS: 38585-74-9 Molecular Formula: C4H5NOS Assay: ≥94% (GC) Chemical Name or Material: Thiazole-5-methanol
Methanol, 99.9%, for HPLC gradient grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: For HPLC gradient grade Assay: ≥99.9% (GC) Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Alfa Aesar™ Methanol, LC-MS Grade, ≥99.8%
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: LC/MS Grade Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 UN Number: UN1230 Beilstein: 1098229 Merck Index: 14,5957
Indole-4-methanol, 97%, Acros Organics™
Melting Point: 46.0° to 50.0°C Water: 1% max. Formula Weight: 147.17 CAS: 1074-85-7 Molecular Formula: C9H9NO Assay: ≥96.0% (GC) Chemical Name or Material: Indole-4-methanol
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Alfa Aesar™
Chemical Name or Material: Methanol CAS: 67-56-1 Name Note: ultrapure Purity Grade: Spectrophotometric grade Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 UN Number: UN1230 Beilstein: 1098229 Merck Index: 14,5957
Methanol (Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.8% Assay Percent Range: ≥99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: OPTIMA™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: MeOH
Methanol, Environmental Grade, >99.8%, Alfa Aesar™
Chemical Name or Material: Methanol Name Note: Environmental Grade CAS: 67-56-1 Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol, 99.9%, for biochemistry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: For biochemistry Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl alcohol
Methanol, ultrapure, Spectrophotometric Grade, >99.8%, Alfa Aesar™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol (Purge and Trap Suitable for Volatile Organic Residue Analysis), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: Purge and trap suitable for volatile organic residue analysis CAS: 67-56-1 Purity Grade: Residue Analysis Grade Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, Semiconductor Grade, 99.9% min., Alfa Aesar™
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Semiconductor Grade Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol (GC Resolv™), Fisher Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: GC Resolv Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, Biograde, >99.8%%, Alfa Aesar™
Chemical Name or Material: Methanol Name Note: Filtered through 0.2μfilter CAS: 67-56-1 Purity Grade: Biochemical Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: Assay: 99.9% min by GC Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: MeOH
Methanol, 99.8%, for electronic use, residue free, ACROS Organics™
Chemical Name or Material: Methanol Name Note: Residue Free CAS: 67-56-1 Purity Grade: For electronic use Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.8%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: ECD Tested, for Pesticide Analysis CAS: 67-56-1 Purity Grade: For residue analysis Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, 99.8%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: for HPLC CAS: 67-56-1 Purity Grade: For HPLC Assay: ≥99.8% (GC) Assay Percent Range: 99.8% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, HPLC Grade, >99.8%, Alfa Aesar™, Ultrapure
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CH4OH Formula Weight: 32.04 MDL Number: MFCD00004595
Methanol (Electronic), Fisher Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Electronic Grade Purity Grade Notes: For use in electronics and circuit board manufacturing Assay: ≥99.8% Assay Percent Range: 99.8% min by GC Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: MeOH
Methanol (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Histological Grade Physical Form: Liquid Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, Optima UHPLCMS, Fisher Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: UHPLC-MS Optima Physical Form: Liquid Assay Percent Range: 99.9% minimum by GC Linear Formula: CH3OH Synonym: MeOH
Methanol (Pesticide), Fisher Chemical
Chemical Name or Material: Methanol CAS: 67-56-1 Purity Grade: Pesticide Grade Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl Alcohol
Methanol, 99.9%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: 99.9% CAS: 67-56-1 Purity Grade: For HPLC Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 MDL Number: MFCD00004595 Synonym: Methyl alcohol
Methanol, ULTRA RESI-ANALYZED, J.T.Baker™
Chemical Name or Material: Methanol CAS: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl alcohol
Methyl Alcohol UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Alcohol CAS: 67-56-1 Molecular Formula: CH3OH Formula Weight: 32.04
Methanol, HPLC Grade, for Anethole Blank, Ricca Chemical
Chemical Name or Material: Methanol Name Note: for Anethole Blank CAS: 67-56-1 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: HPLC Grade Physical Form: Liquid
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methanol Name Note: Peroxide-free CAS: 67-56-1 Purity Grade: Sequencing Grade Purity Grade Notes: Peroxides: ≤0001% Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: ≥99.9 % Molecular Formula: CH4O Linear Formula: CH3OH Formula Weight: 32.04 Synonym: Methyl Alcohol
  spinner