Application Specific Methylene Chloride
Dichloromethane, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: 99.9% CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For HPLC Assay: ≥99.9% Molecular Formula: C H2 Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, for residue analysis, for anal.of polyarom.hydrocarb., stab.with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For residue analysis Assay: ≥99.9% (GC) Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene chloride
Methylene Chloride (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Methylene Chloride (Not Stabilized/HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Not Stabilized CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Physical Form: Liquid Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Dichloromethane, 99.8%, for HPLC, stabilized with amylene, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Stabilized CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For HPLC Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (Stabilized/Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: Stabilized CAS: 75-09-2 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Dichloromethane, 99.8%, for HPLC, stabilized with methanol, ACROS Organics™
Chemical Name or Material: Dichloromethane Name Note: Stabilized CAS: 75-09-2 CAS: 67-56-1 Purity Grade: HPLC Grade Assay: ≥99.8% Assay Percent Range: 99.80% Molecular Formula: CH2Cl2 Linear Formula: CHCl Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: Stabilized with Amylene CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For peptide synthesis Assay: ≥99.9% Assay Percent Range: 99.9% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane, for residue and pestic. anal., stab. with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane Name Note: ECD Tested, For Pesticide analysis CAS: 75-09-2 CAS: 513-35-9 Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Methylene Chloride (With Cyclohexene Preservative/HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride Name Note: With cyclohexene preservative CAS: 75-09-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Dichloromethane, 99+%, extra pure, stabilized with ethanol, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 64-17-5 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: CH2Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Methylene Chloride (Pesticide), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: Pesticide Grade Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM Acidity: Pass Test (USP) Additional Information: Vapor Pressure: Negligible Conductivity: 20 μS/cm max. (EP/BP)
Methylene Chloride (GC Resolv™), Fisher Chemical
Chemical Name or Material: Methylene Chloride CAS: 75-09-2 Purity Grade: GC Resolv Purity Grade Notes: High purity solvent for sample prep prior to trace analysis by GC and/or GC/MS Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: DCM
Dichloromethane, for residue analysis, ACROS Organics™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 CAS: 513-35-9 Purity Grade: For residue analysis Molecular Formula: C H2 Cl2 Linear Formula: CH2Cl2 Formula Weight: 84.93 MDL Number: MFCD00000881 Synonym: Methylene chloride
Dichloromethane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane For gas chromatography ECD and FID SupraSolv™, EMD Millipore
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 UN Number: 1593 Synonym: DCM, Methylene dichloride
Dichloromethane, Stabilized, OmniSolv™, 99.9%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane, Stabilized, 99.9% CAS: 75-09-2 Formula Weight: 84.93
Dichloromethane, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Linear Formula: CH2Cl2 Formula Weight: 84.93
Dichloromethane, Anhydrous, 99.8% min., DriSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane, Anhydrous Name Note: DriSolv™ CAS: 75-09-2 Physical Form: Liquid Assay Percent Range: 99.8%, min. Molecular Formula: CH2Cl2 Linear Formula: K2HPO4 Formula Weight: 174.18 Synonym: Potassium phosphate dibasic
Dichloromethane For spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 UN Number: 1593 Synonym: DCM, Methylene dichloride
Dichloromethane, 99.8% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Name Note: For HPLC, ACS analysis Molecular Formula: CH2Cl2 Formula Weight: 84.93 Synonym: Methylene Chloride
Dichloromethane For gas chromatography MS SupraSolv™, EMD Millipore
Chemical Name or Material: Dichloromethane CAS: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 UN Number: 1593 Synonym: DCM, Methylene dichloride
Dichloromethane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
Additional Information: for pesticide residue analysis Beilstein: 1730800 CAS: 75-09-2 Chemical Name or Material: Dichloromethane Density: 1.325g/mL at 25°C Assay: ≥99.8% (GC) Formula Weight: 84.93g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.22 Health Hazard 2: P260-P280-P305 + P351 + P338 Ignition Point: 1223°F MDL Number: MFCD00000881 RTECS Number: PA8050000 UN Number: UN1593 Vapor Density: 2.9 (vs air) Vapor Pressure: 24.45 psi (55°C), 6.83 psi (20°C)
Dichloromethane CHROMASOLV™, for HPLC, ≥99.8%, contains amylene as stabilizer, Honeywell Riedel-de Haën™
Beilstein: 1730800 CAS: 75-09-2 Chemical Name or Material: Dichloromethane Density: 1.325g/mL at 25°C Assay: ≥99.8% Boiling Point: 39.8 and 40°C Color: APHA: ≤10 Formula Weight: 84.93g/mol Melting Point: −97°C Purity Grade: CHROMASOLV™ EINECS Number: 200-838-9 Health Hazard 2: P261-P281-P305 + P351 + P338 Health Hazard 3: H315-H319-H335-H336-H351-H373 Linear Formula: CH2Cl2 MDL Number: MFCD00000881 Molecular Formula: CH2Cl2 Name Note: for HPLC, contains amylene as stabilizer Refractive Index: n20/D 1.424 RTECS Number: PA8050000 Synonym: Methylene chloride Vapor Density: 2.9 (vs. air) Vapor Pressure: 6.83 psi (20°C)
  spinner