Application Specific Toluene
Toluene, Spectrophotometric Grade, 99.7+%, Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Spectrophotometric grade Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 UN Number: UN1294 Beilstein: 635760 Merck Index: 14,9529
Toluene, 99.85%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: For HPLC Assay: ≥99.8% Assay Percent Range: 99.85% Molecular Formula: C7 H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: Also meets ACS specifications CAS: 108-88-3 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.8% Assay Percent Range: 99.8% min by GC Molecular Formula: C7H8 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, Spectrophotometric Grade, >99.7%, Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Toluene, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: 99+% CAS: 108-88-3 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7 H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, 99.7% min, HPLC Grade, Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99.7% min. Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Toluene, 99.85%, for residue analysis, for pesticide residue analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: For Pesticide Residue Analysis CAS: 108-88-3 Purity Grade: For residue analysis Assay: ≥99.80% (GC) Assay Percent Range: 99.85% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, Environmental Grade, >99.8%, Alfa Aesar™
Chemical Name or Material: Toluene Name Note: Submicron filtered CAS: 108-88-3 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Alfa Aesar™ Toluene, anhydrous, 99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles
Chemical Name or Material: Toluene CAS: 108-88-3 Name Note: Anhydrous, over molecular sieves Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 UN Number: UN1294 Beilstein: 635760 Merck Index: 14,9529
Toluene, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Histological Grade Assay: ≥99.5% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, HPLC Grade, 99.7% min, Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: HPLC Grade Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 UN Number: UN1294 Beilstein: 635760 Merck Index: 14,9529
Toluene, Semiconductor Grade, 99% min., Alfa Aesar™
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Semiconductor Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512
Toluene, 99.85%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: Extra Dry CAS: 108-88-3 Purity Grade: Extra Dry Assay: ≥99.8% (GC) Assay Percent Range: 99.85% Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Toluene CAS: 108-88-3 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene Name Note: 99.8% CAS: 108-88-3 Purity Grade: For HPLC Assay: ≥99.8% Molecular Formula: C7 H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 MDL Number: MFCD00008512 Synonym: Methylbenzene
Toluene, OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Molecular Formula: C7H8 Linear Formula: C6H5CH3 Formula Weight: 92.14 Synonym: Methylbenzene
Toluene For gas chromatography ECD and FID SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 UN Number: 1294 Synonym: Methylbenzene
Toluene For gas chromatography MS SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Toluene CAS: 108-88-3 Molecular Formula: C7H8 Formula Weight: 92.14 UN Number: 1294 Synonym: Methylbenzene
Toluene, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
Additional Information: for pesticide residue analysis Beilstein: 635760 CAS: 108-88-3 Chemical Name or Material: Toluene Density: 0.865g/mL at 25°C Assay: ≥99.7% (GC) Color: Colorless Formula Weight: 92.14g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.07 Flash Point: 4°C (39.2°F) Ignition Point: 997°F Linear Formula: C6H5CH3 MDL Number: MFCD00008512 UN Number: UN1294 Vapor Density: 3.2 (vs air) Vapor Pressure: 22 mmHg (20°C), 26 mmHg (25°C)
Toluene ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Toluene CAS: 108-88-3 Molecular Formula: C6H5CH3 Formula Weight: 92.14
  spinner