Arginine -Arg and Derivatives
NG-Nitro-L-arginine-methyleester hydrochloride, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N-ω-Nitro-L-arginine-methyl ester hydrochloride CAS: 51298-62-5 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C7H15N5O4·HCl Formula Weight: 269.7 MDL Number: MFCD00039052 Synonym: H-Arg(NOZ)-OMe.HCl
Nalpha-Benzoyl-L-arginine ethyleester hydrochloride, ≥99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Nα-Benzoyl-L-arginine ethyl ester hydrochloride CAS: 272320 CAS Min %: 99.0 CAS Max %: 100.0 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C15H22N4O3·HCl Linear Formula: H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl Formula Weight: 342.83 MDL Number: MFCD00012579 Synonym: BAEE
L(+)-Arginine, 98+%, ACROS Organics™
Chemical Name or Material: L(+)-Arginine Name Note: 98+% CAS: 74-79-3 Assay: ≥98.0% (on dried substance) Assay Percent Range: 98+% Molecular Formula: C6H14N4O2 Linear Formula: H2NC(=NH)NH(CH2)3CH(NH2)CO2H Formula Weight: 174.2 MDL Number: MFCD00002635 Synonym: Arg
L-Arginine, 98+%, Alfa Aesar
Chemical Name or Material: L-Arginine Assay Percent Range: 98+% Molecular Formula: C6H14N4O2 Formula Weight: 174.2
Arginine Products for Science Education
Chemical Name or Material: L-Arginine CAS: 74-79-3 Linear Formula: C6H14N4O2 Formula Weight: 174.12 Synonym: Arginine
N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride, 98%, ACROS Organics™
Chemical Name or Material: N-α-Benzoyl-DL-arginine-4-nitroanilide hydrochloride CAS: 911-77-3 CAS Min %: 97.5 CAS Max %: 100.0 Assay: ≥97.5% (HPLC) Assay Percent Range: 98% Molecular Formula: C19H22N6O4·HCl Formula Weight: 434.89 MDL Number: MFCD00012846 Synonym: BANI
NG-Monomethyl-L-arginine monoacetate, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N-ω-Monomethyl-L-arginine monoacetate CAS: 53308-83-1 CAS Min %: 98.5 CAS Max %: 100.0 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C9H20N4O4 Formula Weight: 248.3 MDL Number: MFCD00069311 Synonym: H-Arg(Me)-OH.AcOH
N2-Methyl-L-arginine, Tocris™
Chemical Name or Material: N2-Methyl-L-arginine Assay Percent Range: >98% Molecular Formula: C7H16N4O2 Formula Weight: 188.23 Synonym: N2-Methyl-L-arginine
L-NNA, Tocris
Chemical Name or Material: L-NNA CAS: 2149-70-4 Assay Percent Range: >99% Molecular Formula: C6H13N5O4 Formula Weight: 219.2 Synonym: NG-Nitro-L-arginine
L-NMMA acetate, Tocris
Chemical Name or Material: L-NMMA acetate Assay Percent Range: >98% Molecular Formula: C7H16N4O2·CH3CO2H Formula Weight: 248.28 Synonym: NG-Monomethyl-L-arginine acetate
L-NAME hydrochloride, Tocris
Chemical Name or Material: L-NAME hydrochloride CAS: 51298-62-5 Assay Percent Range: >98% Molecular Formula: C7H15N5O4·HCl Formula Weight: 269.69 Synonym: NG-Nitro-L-arginine methyl ester hydrochloride
L-Arginine, Tocris
Chemical Name or Material: L-Arginine CAS: 74-79-3 Assay Percent Range: >98% Molecular Formula: C6H14N4O2 Formula Weight: 174.2
Nω-Propyl-L-arginine hydrochloride, Tocris
Chemical Name or Material: Nω-Propyl-L-arginine hydrochloride Assay Percent Range: >98% Molecular Formula: C9H20N4O2·HCl Formula Weight: 252.74 Synonym: N5-[Imino(propylamino)methyl]-L-ornithine hydrochloride
L-Arginine, Free Base, Fisher BioReagents
Chemical Name or Material: L-Arginine, Free Base CAS: 74-79-3 Physical Form: Solid Assay Percent Range: ≥99 % Molecular Formula: C6H14N4O2 Formula Weight: 174.12 Synonym: (S)-2-amino-5-[(aminoiminomethyl)amino]pentanoic acid
L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Arginine Hydrochloride CAS: 1119-34-2 Physical Form: Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C6H14N4O2ClH Formula Weight: 210.58
L-Arginine (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Arginine CAS: 74-79-3 Physical Form: Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C6H14N4O2 Formula Weight: 174.2
DL-arginine hydrochloride, MP Biomedicals™
Chemical Name or Material: DL-Arginine CAS: 32042-43-6 Physical Form: Crystalline Molecular Formula: C6H14N4O2.HCl Formula Weight: 210.7 Synonym: 1-Hydrochloride arginine
L-Arginine, 99%, MP Biomedicals™
Chemical Name or Material: L-Arginine free base CAS: 74-79-3 Physical Form: Powder Assay Percent Range: 99% Molecular Formula: C6H14N4O2 Formula Weight: 174.2 Synonym: (S)-2-Amino-5-guanidinopentanoic acid
L-Arginine [Crystalline], MP Biomedicals
Chemical Name or Material: L-Arginine Name Note: Hydrochloride CAS: 1119-34-2 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C6H14N4O2•HCl Formula Weight: 210.7
N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride, MP Biomedicals™
Chemical Name or Material: N-ω-Nitro-L-Arginine Methyl Ester Hydrochloride CAS: 51298-62-5 Physical Form: Powder Molecular Formula: C7H15N5O4·HCl Formula Weight: 269.7 MDL Number: MFCD00039052
L-Arginine, 99%, [Free Base], MP Biomedicals
Chemical Name or Material: L-Arginine, 99% Name Note: Free Base CAS: 74-79-3 Purity Grade: Cell Culture Reagent Grade Molecular Formula: C6H14N4O2 Formula Weight: 174.2
N-acetyl-L-arginine dihydrate, >99%, MP Biomedicals™
Chemical Name or Material: N-acetyl-L-arginine dihydrate CAS: 155-84-0 Physical Form: Powder Assay Percent Range: >99% Molecular Formula: C8H16N4O3·2H2O Formula Weight: 252.27 MDL Number: MFCD00019733 Synonym: 2-Acetamido-5-Guanidinovaleric acid
L-Arginine, Hydrochloride, MP Biomedicals™
Chemical Name or Material: L-Arginine Hydrochloride CAS: 74-79-3 Physical Form: Crystalline Powder Assay Percent Range: 0.992 Molecular Formula: C6H14N4O2 Formula Weight: 174.2 Synonym: Arg
L-Arginine, Hydrochloride, MP Biomedicals
Chemical Name or Material: L-Arginine, Hydrochloride CAS: 1119-34-2 Molecular Formula: C6H14N4O2·ClH Formula Weight: 210.7 Synonym: Argamine
L-arginine methyl ester dihydrochloride, 98%, ACROS Organics™
Chemical Name or Material: L-Arginine methyl ester dihydrochloride Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C7H16N4O2·2HCl Linear Formula: NH2C(NH)N(H)(CH2)3CH(NH2)C(O)OCH3·2HCl Formula Weight: 261.15 MDL Number: MFCD00038948
  spinner