Asparagine -Asn and Derivatives
L-Aspartic acid monosodium salt monohydrate, 97%, Acros Organics
Chemical Name or Material: L-Aspartic acid monosodium salt monohydrate Assay Percent Range: 97% Molecular Formula: C4H6NNaO4·H2O Formula Weight: 173.1
DL-Asparagine monohydrate, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: DL-Asparagine monohydrate Name Note: 98% CAS: 3130-87-8 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H8N2O3·H2O Linear Formula: H2NCOCH2CH(NH2)CO2H·H2O Formula Weight: 150.14 MDL Number: MFCD00151039 Synonym: DL-2-Aminosuccinamic acid
L(+)-Asparagine monohydrate, Acros Organics
Chemical Name or Material: L(+)-Asparagine monohydrate CAS: 5794-13-8 Molecular Formula: C4H8N2O3·H2O Formula Weight: 150.14 Synonym: L-2-Aminosuccinamic acid
L-Asparagine, 99%, ACROS Organics™
Chemical Name or Material: L-Asparagine Assay: ≥98.5% (on dry substance) Assay Percent Range: 99% Molecular Formula: C4H8N2O3 Linear Formula: H2NCOCH2CH(NH2)CO2H Formula Weight: 132.12 MDL Number: MFCD00064401
L+-Asparagine monohydrate, 99%, ACROS Organics™
Chemical Name or Material: L(+)-Asparagine monohydrate Name Note: 99% CAS: 5794-13-8 Assay: 98.5 to 101.5% (on dried substance) Assay Percent Range: 99% Molecular Formula: C4H8N2O3·H2O Linear Formula: H2NCOCH2CH(NH2)CO2H·H2O Formula Weight: 150.14 MDL Number: MFCD00151038 Synonym: L-2-Aminosuccinamic acid
L-Asparagine Products for Science Education
Chemical Name or Material: L-Asparagine CAS: 5794-13-8 Physical Form: Powder Linear Formula: C4H8N2O3·H2O Formula Weight: 150.14 Synonym: Aspartic Acid
L-Aspartic Acid Products for Science Education
Chemical Name or Material: L-Aspartic Acid CAS: 56-84-8 Linear Formula: C4H7NO4 Formula Weight: 133.05 Synonym: Aspartic Acid
WAY 213613, Tocris
Chemical Name or Material: WAY 213613 Assay Percent Range: >99% Molecular Formula: C16H13BrF2N2O4 Formula Weight: 415.19 Synonym: N-[4-(2-Bromo-4, 5-difluorophenoxy)phenyl]-L-asparagine
2,4-Dihydroxyphenylacetyl-L-asparagine, Tocris
Chemical Name or Material: 2, 4-Dihydroxyphenylacetyl-L-asparagine CAS: 111872-98-1 Assay Percent Range: >98% Molecular Formula: C12H14N2O6 Formula Weight: 282.25
L-Asparagine, 99% [Anhydrous], MP Biomedicals
Chemical Name or Material: L-Asparagine, 99% [Anhydrous] CAS: 70-47-3 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C4H8N2O3 Formula Weight: 132.1
D-Asparagine monhydrate, 100.5%, MP Biomedicals™
Chemical Name or Material: D-Asparagine monhydrate CAS: 5794-24-1 Physical Form: Crystalline Assay Percent Range: 1.005 Molecular Formula: C4H8N2O3 · H2O Formula Weight: 150.1 Synonym: (R)-(-)-2-Aminosuccinamic acid
L-Asparagine Anhydrous MP Biomedicals
Chemical Name or Material: L-Asparagine Name Note: Anhydrous CAS: 70-47-3 Molecular Formula: C4H8N2O3 Formula Weight: 132.1
L-Asparagine anhydrous, 100%, MP Biomedicals™
Chemical Name or Material: L-Asparagine CAS: 70-47-3 Physical Form: Powder Assay Percent Range: 1 Molecular Formula: C4H8N2O3 Formula Weight: 132.1 Synonym: (S)-2-Aminosuccinic acid 4-amide
L-Asparagine Monohydrate (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Asparagine Monohydrate CAS: 5794-13-8 Physical Form: Powder/Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C4H8N2O3 · H2O Formula Weight: 150.13
  spinner