Aspartic Acid -Asp and Derivatives
BOC-L-Aspartic acid 4-benzylester, 99+%, ACROS Organics™
Chemical Name or Material: BOC-L-Aspartic acid 4-benzylester Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C16H21NO6 Linear Formula: C6H5CH2OCOCH2CH(COOH)nHCOOC(CH3)3 Formula Weight: 323.34 MDL Number: MFCD00065564 Synonym: N-(tert-Butoxycarbonyl)-L-aspartic acid 4-benzylester
L-aspartic acid dimethyl ester hydrochloride, 99%, ACROS Organics™
Chemical Name or Material: L-Aspartic acid dimethyl ester hydrochloride Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H11NO4·HCl Linear Formula: CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl Formula Weight: 197.62 MDL Number: MFCD00038878
D(-)-Aspartic acid, +99%, ACROS Organics™
Chemical Name or Material: D(-)-Aspartic acid Name Note: 99+% CAS: 1783-96-6 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H7NO4 Linear Formula: HO2CCH2CH(NH2)CO2H Formula Weight: 133.1 MDL Number: MFCD00063081 Synonym: D-2-Aminobutanedioic acid
BOC-L-Aspartic acid 4-methyleester, 98%, ACROS Organics™
Chemical Name or Material: BOC-L-Aspartic acid 4-methyleester Assay: ≥97.5% (Acid) Assay Percent Range: 98% Molecular Formula: C10H17NO6 Formula Weight: 247.25 Synonym: BOC-Asp(OMe)-OH
L(+)-Aspartic Acid +98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L(+)-Aspartic acid Name Note: 98+% CAS: 56-84-8 Assay: ≥98.0% (on dried substance) Assay Percent Range: 98+% Molecular Formula: C4H7NO4 Linear Formula: HO2CCH2CH(NH2)CO2H Formula Weight: 133.1 MDL Number: MFCD00002616 Synonym: L-2-Aminobutanedioic acid
DL-Aspartic Acid +99%, ACROS Organics™
Chemical Name or Material: DL-Aspartic acid Name Note: 99+% CAS: 617-45-8 Assay: 99.0 to 101.0% (Acid) Assay Percent Range: 99+% Molecular Formula: C4H7NO4 Linear Formula: HO2CCH2CH(NH2)CO2H Formula Weight: 133.1 MDL Number: MFCD00063083 Synonym: DL-2-Aminobutanedioic acid
DL-TBOA, Tocris
Chemical Name or Material: DL-TBOA Assay Percent Range: >98% Molecular Formula: C11H13NO5 Formula Weight: 239.23 Synonym: DL-threo-β-Benzyloxyaspartic acid
NMDA, Tocris
Chemical Name or Material: NMDA Assay Percent Range: >99% Molecular Formula: C5H9NO4 Formula Weight: 147.13 Synonym: N-Methyl-D-aspartic acid
TFB-TBOA, Tocris
Chemical Name or Material: TFB-TBOA CAS: 480439-73-4 Assay Percent Range: >98% Molecular Formula: C19H17F3N2O6 Formula Weight: 426.35 Synonym: (3S)-3-[[3-[[4-(Trifluoromethyl)benzoyl]amino]phenyl]methoxy]-L-aspartic acid
L-Aspartic acid, Tocris
Chemical Name or Material: L-Aspartic acid CAS: 56-84-8 Assay Percent Range: >98% Molecular Formula: C4H7NO4 Formula Weight: 133.1 Synonym: L-Aminosuccinic acid
L-Aspartic Acid (White Crystals or Crystalline Powder), Fisher BioReagents
Chemical Name or Material: L-Aspartic Acid CAS: 56-84-8 Physical Form: Solid Assay: 98.5 to 101.0% Assay Percent Range: 98.5 to 101.0 % Molecular Formula: C4H7NO4 Formula Weight: 133.05
L-(-)-threo-3-Hydroxyaspartic acid, Tocris
Chemical Name or Material: L-(-)-threo-3-Hydroxyaspartic acid CAS: 7298-99-9 Assay Percent Range: >97% Molecular Formula: C4H7NO5 Formula Weight: 149.1
L-Aspartate de magnesium salt, MP Biomedicals™
Chemical Name or Material: L-Aspartate de magnesium salt CAS: 18962-61-3 Physical Form: Crystalline Powder Molecular Formula: C8H12N2O8Mg Formula Weight: 288.5 (Anhydrous) Synonym: Magnesium L-aspartate
DL-Aspartic acid, 99+%, MP Biomedicals™
Chemical Name or Material: DL-Aspartic acid CAS: 617-45-8 Physical Form: Crystalline Assay Percent Range: 99+% Molecular Formula: C4H7NO4 Formula Weight: 133.1 Synonym: DL-Aminosuccinic Acid
L-Aspartic Acid, 99+% MP Biomedicals
Chemical Name or Material: L-Aspartic Acid, 99+% CAS: 56-84-8 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C4H7NO4 Formula Weight: 133.1
D-Aspartic Acid, MP Biomedicals™
Chemical Name or Material: D-Aspartic Acid CAS: 1783-96-6 Physical Form: Crystalline Powder Molecular Formula: C4H7NO4 Formula Weight: 133.11 MDL Number: MFCD00063081 Synonym: (R)-(-)-Aminosuccinic acid
  spinner