Atomic Absorption Standards
Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, ACROS Organics™
Chemical Name or Material: Copper standard solution Name Note: 1 mg/ml Cu in 2% HNO3 CAS: 7440-50-8 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Cu Formula Weight: 63.55 MDL Number: MFCD00010965
Vanadium standard solution, for AAS, 1mg/mL V(3+) in 2% HNO3, ACROS Organics™
Chemical Name or Material: Vanadium standard solution Name Note: 1 mg/ml V in 2% HNO3 CAS: 7440-62-2 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: V Formula Weight: 50.94 MDL Number: MFCD00011453
Lead standard solution, for AAS, 1mg/mL Pb in 2% HNO3, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Lead standard solution Name Note: 1 mg/ml Pb in 2% HNO3 CAS: 10099-74-8 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Pb Formula Weight: 331.21 MDL Number: MFCD00134050
Selenium standard solution, for AAS, 1mg/mL Se in 10% HCl, ACROS Organics™
Chemical Name or Material: Selenium standard solution Name Note: 1 mg/ml Se in 10% HCl CAS: 7782-49-2 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Se Formula Weight: 78.96 MDL Number: MFCD00134090
Manganese standard solution, for AAS, 1mg/mL Mn in 2% HNO3, ACROS Organics™
Chemical Name or Material: Manganese standard solution Name Note: 1 mg/ml Mn in 2% HNO3 CAS: 7439-96-5 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Mn Formula Weight: 54.94 MDL Number: MFCD00011111
Aluminium standard solution, for AAS, 1mg/mL Al in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Aluminium standard solution Name Note: 1 mg/ml Al in 0.5N HNO3 CAS: 7784-27-2 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: AlN3O9·9H2O Formula Weight: 375.13 MDL Number: MFCD00134029
Chromium(III) standard solution, for AAS, 1 mg/ml Cr in 2% HCl, ACROS Organics™
Chemical Name or Material: Chromium(III) standard solution Name Note: 1 mg/ml Cr in 2% HCl CAS: 7440-47-3 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Cr Formula Weight: 52 MDL Number: MFCD00143970
Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, ACROS Organics™
Chemical Name or Material: Gold standard solution Name Note: 1 mg/ml Au in 0.5N HCl CAS: 7440-57-5 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Au Formula Weight: 196.97 MDL Number: MFCD00003436
Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH, ACROS Organics™
Chemical Name or Material: Silicon standard solution Name Note: 1 mg/ml Si in 2% NaOH CAS: 7440-21-3 Purity Grade: For AAS Molecular Formula: Si Formula Weight: 28.09 MDL Number: MFCD00085311
Zinc standard solution, for AAS, 1mg/mL Zn in 2% HCl, ACROS Organics™
Chemical Name or Material: Zinc standard solution Name Note: 1 mg/ml Zn in 2% HCl CAS: 7440-66-6 CAS: 7647-01-0 CAS: 7732-18-5 CAS: 7646-85-7 Purity Grade: For AAS Molecular Formula: Zn Formula Weight: 65.38 MDL Number: MFCD00011291
Arsenic standard solution, for AAS, 1mg/mL As in 2% KOH, ACROS Organics™
Chemical Name or Material: Arsenic standard solution Name Note: 1 mg/ml As in 2% KOH CAS: 7440-38-2 CAS: 1310-58-3 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: As Formula Weight: 74.92 MDL Number: MFCD00085309
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, ACROS Organics™
Chemical Name or Material: Cobalt standard solution Name Note: 1 mg/ml Co in 2% HNO3 CAS: 7697-37-2 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Co Formula Weight: 58.93 MDL Number: MFCD00010935
Mercury standard solution, for AAS, solution, 1 mg/ml Hg in 10% HNO3, ACROS Organics™
Chemical Name or Material: Mercury standard solution Name Note: 1 mg/ml Hg in 10% HNO3 CAS: 7783-34-8 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Hg Formula Weight: 342.62 MDL Number: MFCD00011035
Nickel standard solution, for AAS, 1 mg/ml Ni in 2% HNO3, ACROS Organics™
Chemical Name or Material: Nickel standard solution Name Note: 1 mg/ml Ni in 2% HNO3 CAS: 7440-02-0 CAS: 7697-37-2 CAS Min %: 1.9 CAS Max %: 2.1 CAS: 13478-00-7 CAS Min %: 96.9 CAS Max %: 97.1 CAS: 7732-18-5 CAS: na Purity Grade: For AAS Molecular Formula: Ni Formula Weight: 58.69 MDL Number: MFCD00011137
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl, ACROS Organics™
Chemical Name or Material: Lithium standard solution Name Note: 1 mg/ml Li in 2% HCl CAS: 7439-93-2 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Li Formula Weight: 6.94 MDL Number: MFCD00134051
Potassium standard solution, for AAS, 1mg/mL K in 2% HNO3, ACROS Organics™
Chemical Name or Material: Potassium standard solution Name Note: 1 mg/ml K in 2% HNO3 CAS: 7440-09-7 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: K Formula Weight: 39.1 MDL Number: MFCD00133776
Iridium standard solution, for AAS, 1 mg/ml Ir in 5% HCl, ACROS Organics™
Chemical Name or Material: Iridium standard solution CAS: 7732-18-5 CAS: 7647-01-0 Purity Grade: For AAS Molecular Formula: Ir
Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Silver standard solution Name Note: 1 mg/ml Ag in 0.5N HNO3 CAS: 7761-88-8 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Ag Formula Weight: 107.87 MDL Number: MFCD00003397
Niobium standard solution, for AAS, 1mg/mL Nb in 5% HF, ACROS Organics™
Chemical Name or Material: Niobium standard solution Name Note: 1 mg/ml Nb in 5% HF CAS: 7440-03-1 CAS: 7664-39-3 CAS Min %: 4.9 CAS Max %: 5.1 CAS: 7732-18-5 CAS Min %: 93.9 CAS Max %: 94.1 Purity Grade: For AAS Molecular Formula: Nb Formula Weight: 92.91 MDL Number: MFCD00011126
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, ACROS Organics™
Chemical Name or Material: Zirconium standard solution Name Note: 1 mg/ml Zr in 5% HF CAS: 7664-39-3 CAS: 7664-39-3 CAS: 7732-18-5 Purity Grade: For AAS MDL Number: MFCD00011303
Iron standard solution, for AAS, 1 mg/ml Fe in 2% HNO3, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Iron standard solution Name Note: 1 mg/ml Fe in 2% HNO3 CAS: 7439-89-6 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Fe Formula Weight: 55.85 MDL Number: MFCD00010999
Magnesium standard solution, for AAS, 1mg/mL Mg in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Magnesium standard solution Name Note: 1 mg/ml Mg in 0.5N HNO3 CAS: 7439-95-4 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Mg Formula Weight: 24.31 MDL Number: MFCD00085308
Platinum standard solution, for AAS, 1mg/mL Pt in 10% HCl, ACROS Organics™
Chemical Name or Material: Platinum standard solution Name Note: 1 mg/ml Pt in 10% HCl CAS: 7647-01-0 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Linear Formula: Pt MDL Number: MFCD00011179
Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF, ACROS Organics™
Chemical Name or Material: Hafnium standard solution Name Note: 1 mg/ml Hf in 5% HF CAS: 7664-39-3 CAS: 7664-39-3 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Hf Formula Weight: 178.49 MDL Number: MFCD00011032
Ruthenium standard solution, for AAS, 1mg/mL Ru in 5% HCl, ACROS Organics™
Chemical Name or Material: Ruthenium standard solution Name Note: 1 mg/ml Ru in 5% HCl CAS: 7440-18-8 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Ru Formula Weight: 101.07 MDL Number: MFCD00011207
Calcium standard solution, for AAS, 1mg/mL Ca in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Calcium standard solution Name Note: 1 mg/ml Ca in 0.5N HNO3 CAS: 7440-70-2 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Ca Formula Weight: 40.08 MDL Number: MFCD00085314
Sodium standard solution, for AAS, 1mg/mL Na in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Sodium standard solution Name Note: 1 mg/ml Na in 0.5N HNO3 CAS: 7631-99-4 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Na Formula Weight: 84.99 MDL Number: MFCD00085307
Palladium standard solution, for AAS, 1 mg/ml Pd in 10% HCl, ACROS Organics™
Chemical Name or Material: Palladium standard solution Name Note: 1 mg/ml Pd in 10% HCl CAS: 7647-01-0 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: For AAS MDL Number: MFCD00011167
Rhenium standard solution, for AAS, 1mg/mL Re in water, ACROS Organics™
Chemical Name or Material: Rhenium standard solution CAS: 13598-65-7 CAS: 7732-18-5 Purity Grade: For AAS MDL Number: MFCD00011195
Cadmium standard solution, for AAS, 1mg/mL Cd in 0.5N HNO3, ACROS Organics™
Chemical Name or Material: Cadmium standard solution Name Note: 1 mg/ml Cd in 0.5N HNO3 CAS: 10022-68-1 CAS: 7697-37-2 CAS: 7732-18-5 Purity Grade: For AAS Molecular Formula: Cd Formula Weight: 308.48 MDL Number: MFCD00010914
  spinner