Azelaic Acid
Azelaic acid, 98%, ACROS Organics™
Chemical Name or Material: Azelaic acid Name Note: 98% CAS: 123-99-9 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C9H16O4 Linear Formula: HOOC(CH2)7COOH Formula Weight: 188.22 MDL Number: MFCD00004432 Synonym: Nonanedioic acid
Azelaic Acid, MP Biomedicals
Chemical Name or Material: Azelaic Acid CAS: 123-99-9 Formula Weight: 188.22
Azelaic Acid, 95%, MP Biomedicals
Chemical Name or Material: Azelaic Acid Name Note: 95% CAS: 123-99-9 Formula Weight: 188.22
  spinner