Barbituric Acid
Barbituric Acid, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Barbituric Acid CAS: 67-52-7 Assay: ≥98% Assay Percent Range: ≥98 % Molecular Formula: C4H4N2O3 Linear Formula: NHCONHCOCH2CO Formula Weight: 128.09 MDL Number: MFCD00006666 Synonym: Malonylurea
Barbituric Acid (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Barbituric Acid CAS: 67-52-7 Purity Grade: Laboratory Grade Molecular Formula: C4H4N2O3 Linear Formula: NHCONHCOCH2CO Formula Weight: 128.09 MDL Number: MFCD00006666 Synonym: Malonylurea
  spinner