Barium -Ba
Barium Pieces, 2.5cm (1 in.) & down, 99.2% (Metals basis), Alfa Aesar™
Chemical Name or Material: Barium CAS: 7440-39-3 Physical Form: Pieces, 2.5cm (1 in.) and down Assay Percent Range: 99.2% (Metals basis) MDL Number: MFCD00134031
Barium Crystalline dendritic solid, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Barium CAS: 7440-39-3 Physical Form: Crystalline Dendritic Solid Assay Percent Range: 99.9% (Metals basis) MDL Number: MFCD00134031
Barium, 99+%, rods, ∽2cm diameter, in mineral oil, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Barium Name Note: 99+% CAS: 7440-39-3 CAS Min %: 99.0 CAS Max %: 100.0 Physical Form: Rod Assay Percent Range: 99+% Molecular Formula: Ba Linear Formula: Ba Formula Weight: 137.34
  spinner