Boric Acid
Boric Acid (Crystalline/NF/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: BP/EP/JP/NF Physical Form: Powder/Solid Assay: 99.5 to 100.5% (NF) Molecular Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Synonym: Borofax
Alfa Aesar™ Boric acid, Puratronic™, 99.9995% (metals basis)
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Puratronic™ Molecular Formula: BH3O3 Linear Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Beilstein: 1697939 Merck Index: 14,1336 Synonym: Boracic Acid
Boric acid, ACS, 99.5% min, Alfa Aesar
In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates
Boric Acid (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Synonym: Borofax
Boric Acid (Crystalline/NF), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: NF Physical Form: Powder/Solid Assay: 99.5 to 100.5% (NF) Molecular Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Synonym: Borofax
Boric Acid Solution with Indicator, Fisher Chemical
Chemical Name or Material: Boric Acid Solution with Indicator CAS: 7732-18-5 CAS Min %: 94.0 CAS Max %: 98.0 CAS: 10043-35-3 CAS Min %: 3.76 CAS Max %: 3.92 CAS: 64-17-5 CAS Min %: 0.2 CAS Max %: 0.3 CAS: 67-56-1 CAS Min %: 0.01 CAS Max %: 0.02 CAS: 67-63-0 CAS Min %: 0.1 CAS Max %: 0.2 CAS: 63451-28-5 CAS Min %: 1.0E-4 CAS Max %: 1.0E-4
Boric Acid (Powder/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Certified ACS Grade Physical Form: Powder/Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Synonym: Borofax
Boric Acid (Laboratory), Fisher Chemical
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Laboratory Grade Molecular Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337 Synonym: Borofax
Boric acid, 99.6%, ACS reagent, ACROS Organics™
Chemical Name or Material: Boric acid CAS: 10043-35-3 Purity Grade: ACS Reagent Grade Assay: ≥99.5% (Acid) Assay Percent Range: 99.6% Molecular Formula: H3BO3 Linear Formula: H3BO3 Formula Weight: 61.83 MDL Number: MFCD00011337
Boric Acid, 4% (w/v) with Bromocresol Green-Methyl Red, for Kjeldahl Analysis, Ricca Chemical
Chemical Name or Material: Boric Acid Name Note: With Bromocresol Green-Methyl Red Indicator CAS: 7732-18-5,10043-35-3, 62625-32-5, 845-10-3 CAS Min %: 94.0 CAS Max %: 98.0 CAS: 10043-35-3 CAS Min %: 3.77 CAS Max %: 3.92 CAS: 62625-32-5 CAS Min %: 9.0E-4 CAS Max %: 9.0E-4 CAS: 845-10-3 CAS Min %: 7.0E-4 CAS Max %: 7.0E-4 Physical Form: Liquid
Boric Acid, 2% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Boric Acid CAS: 7732-18-5 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 10043-35-3 CAS Min %: 1.92 CAS Max %: 2.0 Physical Form: Liquid
Boric Acid, 4% (w/v) Aq Soln, Ricca Chemical
Chemical Name or Material: Boric Acid Solution Name Note: 4%(w/v) Aqueous Solution CAS: 7732-18-5 CAS Min %: 94.0 CAS Max %: 98.0 CAS: 10043-35-3 CAS Min %: 3.77 CAS Max %: 3.93
Boric Acid ACS AR Granular, Macron Fine Chemicals™
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: ACS Grade Physical Form: Granular Molecular Formula: H3BO3 Formula Weight: 61.83
Boric Acid (Crystalline/Electrophoresis), Fisher BioReagents
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Electrophoresis Grade Purity Grade Notes: DNase-Free Physical Form: Powder/Solid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: H3BO3 Formula Weight: 61.83
Boric Acid, 4% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Boric Acid CAS: 7732-18-5 CAS Min %: 94.0 CAS Max %: 98.0 CAS: 10043-35-3 CAS Min %: 3.77 CAS Max %: 3.93 Physical Form: Liquid
Boric Acid ACS Reagent Grade, Ricca Chemical
Chemical Name or Material: Boric Acid CAS: 10043-35-3 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade
Boric Acid NF GenAR Granular, Macron Fine Chemicals™
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: NF Physical Form: Granular Molecular Formula: H3BO3 Formula Weight: 61.83
Boric Acid Aqueous Solution, 3% (w/v), w/Bromocresol Green-Methyl Red Indicator, Ricca Chemical
CAS: 10043-35-3 CAS: 62625-32-5 CAS: 7732-18-5 CAS: 845-10-3 Physical Form: Solution Assay: 3% Assay Percent Range: 3% (w/v)
Boric acid, 100.28%, MP Biomedicals™
Chemical Name or Material: Boric acid CAS: 10043-35-3 Physical Form: Crystals Assay Percent Range: 1.0028 Molecular Formula: BH3O3 Formula Weight: 61.83 Synonym: Boracic acid
Boric Acid, 99.5% (ACS Guaranteed Reagent), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Boric Acid, 99.5% Purity Grade: ACS Grade Physical Form: Crystalline Formula Weight: 61.83
Boric Acid, 99.5 to 100.5%, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Boric Acid Name Note: For ACS analysis CAS: 10043-35-3 Physical Form: Powder Assay Percent Range: 99.5 -100.5% Molecular Formula: H3BO3 Formula Weight: 61.83 Synonym: Trihydroxidoboron
Boric Acid, Molecular Biology Grade, Hoefer™
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Purity Grade: Molecular Biology Grade Physical Form: Crystalline Powder Assay Percent Notes: ≥99.5% Linear Formula: H3BO3 Formula Weight: 61.83
Boric Acid Products for Science Education
Chemical Name or Material: Boric Acid CAS: 10043-35-3 Linear Formula: H3BO3 Formula Weight: 61.83
  spinner