Butanol -tert-
tert-Butanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: tert-Butanol CAS: 75-65-0 Purity Grade: Certified Grade Molecular Formula: C4H10O Linear Formula: (CH3)3COH Formula Weight: 74.12 MDL Number: MFCD00004464 Synonym: 2-Methyl-2-propanol
tert-Butanol, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: tert-Butanol CAS: 75-65-0 Purity Grade: For analysis Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3)3COH Formula Weight: 74.12 Synonym: 2-Methyl-2-propanol
tert-Butanol, 99.5%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: tert-Butanol CAS: 75-65-0 Purity Grade: Extra Pure Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: C4H10O Linear Formula: (CH3)3COH Formula Weight: 74.12 MDL Number: MFCD00004464 Synonym: 2-Methyl-2-propanol
tert-Butanol, ACROS Organics™
Chemical Name or Material: tert-Butanol Name Note: For analysis ACS CAS: 75-65-0 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Molecular Formula: C4H10O Linear Formula: (CH3)3COH Formula Weight: 74.12 MDL Number: MFCD00004464 Synonym: 2-Methyl-2-propanol
tert-Butanol Products for Science Education
CAS: 75-65-0 Linear Formula: (CH3)3COH Formula Weight: 74.12 Synonym: 2-Methyl-2-Propanol
tert-Butyl Alcohol ACS AR, Macron Fine Chemicals™
Chemical Name or Material: tert-Butyl Alcohol CAS: 75-65-0 Purity Grade: ACS Reagent Grade Molecular Formula: (CH3)3COH Formula Weight: 74.12
  spinner