Butanone -2-
2-Butanone, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone CAS: 78-93-3 Purity Grade: For HPLC Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
Methyl Ethyl Ketone (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl Ethyl Ketone CAS: 78-93-3 Purity Grade: Certified ACS Reagent Grade Assay: ≥99% Assay Percent Range: ≥99% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: 2-Butanone
2-Butanone, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone Name Note: Extra Dry CAS: 78-93-3 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
Methyl Ethyl Ketone (Technical), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl Ethyl Ketone CAS: 78-93-3 Purity Grade: Technical Grade Molecular Formula: C4H8O Linear Formula: CH3COC2H5 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: 2-Butanone
2-Butanone, for analysis ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone CAS: 78-93-3 Purity Grade: ACS Reagent Grade Assay: ≥99.0% (GC) Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
2-Butanone, +99%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone CAS: 78-93-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
2-Butanone, 99+%, for electronic use, residue free, ACROS Organics™
Chemical Name or Material: 2-Butanone Name Note: Residue Free CAS: 78-93-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For electronic use Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
2-Butanone, +99%, Extra Pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone CAS: 78-93-3 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O Linear Formula: C2H5COCH3 Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
2-Butanone, ACS, 99+%, Alfa Aesar™
Chemical Name or Material: 2-Butanone CAS: 78-93-3 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C4H8O Formula Weight: 72.11 MDL Number: MFCD00011648 Synonym: MEK
Methyl Ethyl Ketone ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Ethyl Ketone CAS: 78-93-3 Purity Grade: ACS Reagent Grade Molecular Formula: CH3COCH2CH3 Formula Weight: 72.11
Methyl Ethyl Ketone ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Methyl Ethyl Ketone CAS: 78-93-3 Molecular Formula: CH3COCH2CH3 Formula Weight: 72.11
Methyl Ethyl Ketone,MP Biomedicals
Chemical Name or Material: Methyl ethyl ketone CAS: 78-93-3 Physical Form: Liquid Molecular Formula: C4H8O Formula Weight: 72.11 Synonym: 2-Butanone
2-Butanone, 99.0% min, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Butanone Name Note: For ACS analysis CAS: 78-93-3 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: CH3COCH2CH3 Linear Formula: CH3COCH2CH3 Formula Weight: 72.11 Synonym: Methyl Ethyl Ketone
Omnisolv Methyl Ethyl Ketone, 99.5% EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Methyl Ethyl Ketone, 99.5% CAS: 78-93-3 Formula Weight: 72.11
2-Butanone (MEK), ≥95%, Ultra Scientific
Chemical Name or Material: 2-Butanone (MEK) CAS: 000078-93-3 CAS Max %: 100.0 Assay Percent Range: >95%
  spinner