Butyric Acid
Butyric acid sodium salt, 99%, ACROS Organics™
Chemical Name or Material: Butyric Acid sodium salt Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C4H7NaO2 Linear Formula: CH3CH2CH2COONa Formula Weight: 110.09 MDL Number: MFCD00002816 Synonym: Sodium butanoate
n-Butyric acid, +99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Butyric Acid CAS: 107-92-6 CAS Min %: 99.0 CAS Max %: 100.0 Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O2 Linear Formula: CH3CH2CH2CO2H Formula Weight: 88.11
Butyric Acid Products for Science Education
Chemical Name or Material: Butyric Acid CAS: 107-92-6 Purity Grade: Laboratory Grade Assay Percent Range: >99% Linear Formula: CH3CH2CH2COOH Formula Weight: 88.11 Synonym: N-Butanoic Acid
  spinner