Cadmium -Cd
Cadmium shot, 99.9999% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Cadmium CAS: 7440-43-9 Physical Form: Shot, 1-3mm (0.04-0.1 in.) Assay Percent Range: 99.9997% (Metals basis) MDL Number: MFCD00010914
Cadmium Metal Products for Science Education
Chemical Name or Material: Cadmium Metal CAS: 7440-43-9 Assay Percent Range: >99% Linear Formula: Cd Formula Weight: 112.4
Cadmium (Laboratory), Fisher Chemical
Chemical Name or Material: Cadmium CAS: 7440-43-9 Purity Grade: Laboratory Grade Physical Form: Solid Molecular Formula: Cd Formula Weight: 112.41 MDL Number: MFCD00010914
Cadmium, Granular, ACROS Organics™
Chemical Name or Material: Cadmium CAS: 7440-43-9 Molecular Formula: Cd Linear Formula: Cd Formula Weight: 112.4 MDL Number: MFCD00010914
Cadmium Granular, For Analysis, Particle size about 3-6mm, EMSURE™, EMD Millipore™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cadmium CAS: 7440-43-9 Molecular Formula: Cd (Hill) Formula Weight: 112.41 UN Number: UN 3288
Cadmium metal Products for Science Education
CAS: 7440-43-9 Physical Form: Solid Linear Formula: Cd Formula Weight: 112.4
  spinner