Calcium -Ca
Calcium shot, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Calcium CAS: 7440-70-2 Physical Form: Shot, Redistilled, 1cm (0.4 in.) &and down Assay Percent Range: 99.5% (Metals basis) MDL Number: MFCD00085314
Calcium granules, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Calcium Name Note: Redistilled CAS: 7440-70-2 Physical Form: Granular Assay Percent Range: 99.5% (Metals basis) MDL Number: MFCD00085314
Calcium granules, -9 mesh, 98.8% (metals basis), Alfa Aesar™
Chemical Name or Material: Calcium CAS: 7440-70-2 Physical Form: Granular Assay Percent Range: 98.8% (Metals basis) MDL Number: MFCD00085314
Calcium ingot, 99% (metals basis), Alfa Aesar
Chemical Name or Material: Calcium CAS: 7440-70-2 Physical Form: Ingot Assay Percent Range: 99% (metals basis) Molecular Formula: Ca Formula Weight: 40.078
Alfa Aesar™ Calcium granules, redistilled, -16 mesh, 99.5% (metals basis), Alfa Aesar™
Chemical Name or Material: Calcium CAS: 7440-70-2 Name Note: Redistilled MDL Number: MFCD00085314
Calcium, 99%, turnings, ACROS Organics™
Chemical Name or Material: Calcium Name Note: 99% CAS: 7440-70-2 Physical Form: Liquid Assay: ≥98.5% (Titrimetry other) Assay Percent Range: 99% Molecular Formula: Ca Linear Formula: Ca Formula Weight: 40.07 MDL Number: MFCD00085314
Calcium Products for Science Education
Chemical Name or Material: Calcium CAS: 7440-70-2 Linear Formula: Ca Formula Weight: 40.08 Synonym: Calcicat
Calcium, 99%, granular, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Calcium Name Note: 99% CAS: 7440-70-2 Assay: ≥98.5% (Ca) Assay Percent Range: 99% Molecular Formula: Ca Linear Formula: Ca Formula Weight: 40.07 MDL Number: MFCD00085314
Calcium turnings, 98% (metals basis), Alfa Aesar
Chemical Name or Material: Calcium CAS: 7440-70-2 Physical Form: Turnings Assay Percent Range: 98% (metals basis) Molecular Formula: Ca Formula Weight: 40.078
Calcium 99.9% (Cont. 0.1mg), MP Biomedicals
Chemical Name or Material: Calcium CAS: 7440-70-2
Calcium, Turnings 99.5%, ACROS Organics™
Chemical Name or Material: Calcium Name Note: 99.5% CAS: 7440-70-2 Assay: ≥99.4% (Ca) Molecular Formula: Ca Linear Formula: Ca Formula Weight: 40.07
  spinner