Camphoric Acid
D(+)-Camphoric acid, 99%, ACROS Organics™
Chemical Name or Material: D(+)-Camphoric acid CAS: 124-83-4 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C10H16O4 Formula Weight: 200.23g/mol MDL Number: MFCD00001375
D-Camphoric Acid, MP Biomedicals
Chemical Name or Material: D-Camphoric Acid CAS: 124-83-4 Formula Weight: 200.23g/mol
  spinner