Cesium -Cs
Cesium bromide, 99.9%, pure, ACROS Organics™
Chemical Name or Material: Cesium bromide Name Note: 99.9% CAS: 7787-69-1 Purity Grade: Extra Pure Assay Percent Range: 99.90% Molecular Formula: BrCs Linear Formula: CsBr Formula Weight: 212.81 MDL Number: MFCD00010954
Cesium nitrate, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium nitrate CAS: 7789-18-6 Physical Form: Crystalline Assay Percent Range: 99.99% (Metal basis) Molecular Formula: CsNO3 Formula Weight: 194.91 MDL Number: MFCD00010963
Cesium, 99.95+%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Cesium Name Note: 99.95+% CAS: 7440-46-2 Purity Grade: Trace Metal Basis Assay: ≥99.950% Assay Percent Range: 99.95+% Molecular Formula: Cs Formula Weight: 132.91 MDL Number: MFCD00134037
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade, ACROS Organics™
Chemical Name or Material: Cesium chloride Name Note: For spectroscopy CAS: 7647-17-8 Purity Grade: Trace Metal Basis Assay: ≥99.99%(CsCl), purity based on the total trace impurities content (typically%gt; 99.995%) Assay Percent Range: 99.99% Molecular Formula: ClCs Linear Formula: CsCl Formula Weight: 168.36 MDL Number: MFCD00010955
Cesium carbonate, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium carbonate CAS: 534-17-8 Physical Form: Powder/Granules Assay Percent Range: 99% (metals basis) Molecular Formula: Cs2CO3 Formula Weight: 325.82 MDL Number: MFCD00010957
Cesium nitrate, 99.99%, (trace metal basis), for analysis, ACROS Organics™
Chemical Name or Material: Cesium Nitrate CAS: 7789-18-6 Purity Grade: Trace Metal Basis Assay Percent Range: 99.99% Molecular Formula: CsNO3 Linear Formula: CsNO3 Formula Weight: 194.91 MDL Number: MFCD00010963
Cesium acetate, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium acetate CAS: 3396-11-0 Physical Form: Crystalline Powder Assay Percent Range: 99% (Metal basis) Molecular Formula: CsOOCCH3 Formula Weight: 191.95 MDL Number: MFCD00013056
Cesium acetate, for analysis, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium acetate CAS: 3396-11-0 Purity Grade: For analysis Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C2H3CsO2 Linear Formula: CH3CO2Cs Formula Weight: 191.96 MDL Number: MFCD00013056
Cesium fluoride, 99% (metals basis)
Chemical Name or Material: Cesium fluoride CAS: 13400-13-0 Molecular Formula: CsF Formula Weight: 151.9 MDL Number: MFCD00010960 UN Number: UN2923
Cesium chloride, 99.999%, (trace metal basis), extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium chloride CAS: 7647-17-8 Purity Grade: Trace Metal Basis Assay Percent Range: 99.999% Molecular Formula: ClCs Linear Formula: CsCl Formula Weight: 168.36
Cesium carbonate, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium carbonate Name Note: 99.5% CAS: 534-17-8 Purity Grade: For analysis Assay: ≥99.4% (by difference) Assay Percent Range: 99.5% Molecular Formula: CCs2O3 Linear Formula: Cs2CO3 Formula Weight: 325.82 MDL Number: MFCD00010957
Cesium carbonate, 99.994% (metals basis), Alfa Aesar™, Puratronic™
Chemical Name or Material: Cesium carbonate CAS: 534-17-8 Physical Form: Powder Assay Percent Range: 99.994% (Metal basis) Molecular Formula: Cs2CO3 Formula Weight: 325.82 MDL Number: MFCD00010957
Cesium chloride, White powder/crystalline/beads, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Cesium chloride CAS: 7647-17-8 Physical Form: Powder/Crystalline/Beads Assay Percent Range: 99.99% (Metals basis) Molecular Formula: CsCl Formula Weight: 168.36 MDL Number: MFCD00010955
Cesium perchlorate 99% , ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium perchlorate Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: ClCsO4 Linear Formula: CsClO4 Formula Weight: 232.36 MDL Number: MFCD00010956
Cesium nitrate, 99.999% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium nitrate CAS: 7789-18-6 Physical Form: Crystalline Assay Percent Range: 99.999% (Metal basis) Molecular Formula: CsNO3 Formula Weight: 194.91 MDL Number: MFCD00010963
Cesium iodide, 99.9%, for analysis, ACROS Organics™
Chemical Name or Material: Cesium iodide Name Note: 99.9% CAS: 7789-17-5 Purity Grade: For analysis Assay: ≥99.85% Assay Percent Range: 99.9% Molecular Formula: CsI Linear Formula: CsI Formula Weight: 259.8 MDL Number: MFCD00010962
Cesium bromide, White powder/crystalline, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Cesium bromide CAS: 7787-69-1 Physical Form: Powder/Crystalline Assay Percent Range: 99.9% (Metals basis) Molecular Formula: BrCs Formula Weight: 212.81 MDL Number: MFCD00010954
Cesium iodide, 99.999%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium iodide CAS: 7789-17-5 Purity Grade: Trace Metal Basis Assay Percent Range: 99.999% Molecular Formula: CsI Linear Formula: CsI Formula Weight: 259.8
Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Cesium formate hydrate Physical Form: Chunks and Powder Assay Percent Range: 99.9% (Metals basis) Molecular Formula: CsOOCH·xH2O Formula Weight: 177.92 MDL Number: MFCD00039103
Cesium chloride, 99+%, pure, ACROS Organics™
Chemical Name or Material: Cesium chloride Name Note: Pure CAS: 7647-17-8 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: ClCs Linear Formula: CsCl Formula Weight: 168.36 MDL Number: MFCD00010955
Cesium tungsten oxide, White powder, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Cesium tungsten oxide CAS: 13587-19-4 Physical Form: Powder Assay Percent Range: 99.9% (Metals basis) Molecular Formula: Cs2O4W Formula Weight: 513.66 MDL Number: MFCD00049438 Synonym: Cesium tungstate
Cesium, Silver-colored solid, 99.8% (Metals basis), Alfa Aesar™
Chemical Name or Material: Cesium CAS: 7440-46-2 Physical Form: Solid Assay Percent Range: 99.8% (Metals basis) MDL Number: MFCD00134037
Cesium chloride, 99.9% (metals basis), Alfa Aesar
Chemical Name or Material: Cesium chloride CAS: 7647-17-8 Physical Form: -20 Mesh Crystalline Assay Percent Range: 99.9% (metals basis) Molecular Formula: CsCl Formula Weight: 168.36
Cesium formate monohydrate, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cesium formate monohydrate Purity Grade: For analysis Assay: ≥99.5% Molecular Formula: CHCsO2·H2O Linear Formula: HCOOCs Formula Weight: 195.94 MDL Number: MFCD00039103
Cesium chloride, 99.999% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium chloride CAS: 7647-17-8 Physical Form: Crystalline Assay Percent Range: 99.999% (Metal basis) Molecular Formula: CsCl Formula Weight: 168.36 MDL Number: MFCD00010955
Cesium carbonate, 99.995%, (trace metal basis), ACROS Organics™
Chemical Name or Material: Cesium carbonate CAS: 534-17-8 Purity Grade: Trace Metal Basis Assay: 81% Cs (typical analysis) Assay Percent Range: 99.995% Molecular Formula: CCs2O3 Linear Formula: Cs2CO3 Formula Weight: 325.82 MDL Number: MFCD00010957
Cesium Sulfate, 99.9% (metals basis), Alfa Aesar
Chemical Name or Material: Cesium sulfate CAS: 10294-54-9 Physical Form: Powder Assay Percent Range: 99.9% (Metal basis) Molecular Formula: Cs2SO4 Formula Weight: 361.87 MDL Number: MFCD00010959
Cesium carbonate, 99.9% (metals basis), Alfa Aesar
Chemical Name or Material: Cesium carbonate CAS: 534-17-8 Physical Form: -20 Mesh Powder Assay Percent Range: 99.9% (metals basis) Molecular Formula: Cs2CO3 Formula Weight: 325.82
Cesium sulfate, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium sulfate CAS: 10294-54-9 Physical Form: Powder Assay Percent Range: 99.99% (Metal basis) Molecular Formula: Cs2SO4 Formula Weight: 361.87 MDL Number: MFCD00010959
Cesium hydroxide hydrate, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Cesium hydroxide hydrate CAS: 12260-45-6 Physical Form: Crystalline Assay Percent Range: 99.9% (Metal basis) Molecular Formula: CsOH·xH2O (15-20% H2O) Formula Weight: 149.91 (Anhydrous) MDL Number: MFCD00010964
  spinner