Chlorobenzene
Chlorobenzene, Spectrophotometric Grade, 99.9%, Alfa Aesar™
Chemical Name or Material: Chlorobenzene CAS: 108-90-7 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530
Chlorobenzene, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chlorobenzene Name Note: 99+% CAS: 108-90-7 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H5Cl Linear Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530
Chlorobenzene, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Chlorobenzene Name Note: Extra Dry CAS: 108-90-7 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C6H5Cl Linear Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530
Chlorobenzene, 99+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chlorobenzene CAS: 108-90-7 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H5Cl Linear Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530
Chlorobenzene, 99.6%, ACS reagent, ACROS Organics™
Chemical Name or Material: Chlorobenzene Name Note: For analysis ACS CAS: 108-90-7 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C6H5Cl Linear Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530
Chlorobenzene (Laboratory), Fisher Chemical
Chemical Name or Material: Chlorobenzene CAS: 108-90-7 Purity Grade: Laboratory Grade Molecular Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530 Synonym: Benzene Chloride
Chlorobenzene (Certified), Fisher Chemical
Chemical Name or Material: Chlorobenzene CAS: 108-90-7 Purity Grade: Certified Grade Molecular Formula: C6H5Cl Formula Weight: 112.56 MDL Number: MFCD00000530 Synonym: Benzene Chloride
Chlorobenzene ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Chlorobenzene CAS: 108-90-7 Purity Grade: ACS Reagent Grade Molecular Formula: C6H5Cl Formula Weight: 112.56
Chlorobenzene, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chlorobenzene Name Note: For Synthesis CAS: 108-90-7 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: C6H5Cl Linear Formula: C6H5Cl Formula Weight: 112.56 Synonym: Phenyl chloride
  spinner