Chromatography Media
GE Healthcare SOURCE™ 30S Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides
GE Healthcare TALON™ Superflow Medium
Offers different selectivity compared to nickel-charged media. GE Healthcare TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.
GE Healthcare MabSelect™ Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Protein A affinity medium designed for capturing MAbs from large sample volumes
GE Healthcare Phenyl Sepharose™ 6 Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Used for capture and intermediate purification requiring low to medium hydrophobicity
GE Healthcare TALON™ Superflow Medium PROMO
Used for purification of histidine-tagged recombinant proteins by IMAC. GE Healthcare Lifescience™ TALON™ Superflow Medium offers different selectivity compared to nickel-charged media
GE Healthcare MabSelect Xtra™ Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Provides high purity capture and high dynamic binding capacity
GE Healthcare His SpinTrap™ TALON™ Available on GSA/VA Contract for Federal Government customers only.
Single-use spin column for high purity purification of histidine-tagged proteins
GE Healthcare SP Sepharose™ XL Available on GSA/VA Contract for Federal Government customers only.
Strong cation exchanger with high loading capacity and chemical stability
GE Healthcare Capto™ Multimodal (MMC) Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Used to solve specific purification challenges at both high and low ionic strengths
GE Healthcare rProtein A Sepharose™ Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Designed for purification of monoclonal and polyclonal antibodies at labs and process scales
GE Healthcare Sepharose™ Fast Flow Cation Exchange Media Available on GSA/VA Contract for Federal Government customers only.
Used for preparative protein separations in both research and industrial applications. GE Healthcare CM Sepharose™ Fast Flow Cation Exchange Media is composed of crosslinked 6% agarose beads, with carboxymethyl (CM) weak cation exchange groups.
GE Healthcare Capto™ adhere ImpRes Multimodal BioProcess™ Chromatography Medium Available on GSA/VA Contract for Federal Government customers only.
For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules
GE Healthcare Sephaorose™ Fast Flow Anion Exchange Media: ANX Available on GSA/VA Contract for Federal Government customers only.
Used for preparative protein separations in both research and industrial applications
GE Healthcare MabSelect Sure™ LX Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Protein A affinity medium with high dynamic binding capacity which improves performance and cuts costs
GE Healthcare nProtein A Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Used for purification of monoclonal and polyclonal antibodies.   GE Healthcare Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.
GE Healthcare Capto™ S Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Designed for capture and intermediate purification of proteins from large feed volumes. GE Heathcare Capto™ S Chromatography Media is a strong cation exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers.
GE Healthcare His GraviTrap™ TALON™ Available on GSA/VA Contract for Federal Government customers only.
Offers a different selectivity compared to nickel-charged media
GE Healthcare rmp Protein A Sepharose™ Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Designed for high-purity separation of monoclonal and polyclonal antibodies
GE Healthcare HIC (Hydrophobic Interaction Chromatography) Media Available on GSA/VA Contract for Federal Government customers only.
Features high chemical stability, properties to increase productivity and epoxy coupling
GE Healthcare Phenyl Sepharose™ High Performance Media Available on GSA/VA Contract for Federal Government customers only.
Designed for intermediate and polishing step purification steps
GE Healthcare Q Sepharose™ XL Available on GSA/VA Contract for Federal Government customers only.
Hydrohphilic base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in elution pool
GE Healthcare Butyl Sepharose™ 4 Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Used for capture and intermediate purification of larger proteins
GE Healthcare Butyl-S Sepharose™ 6 Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
The least hydrophobic medium in the Sepharose Fast Flow hydrophobic interaction chromatography (HIC) range and is particularly useful during the initial stages of purification process
GE Healthcare NHS-Activated Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Use for coupling amino-containing smaller proteins and peptides.  GE Healthcare Life Sciences™ NHS-Activated Sepharose™ 4 Fast Flow Affinity Media yields coupled products that are very stable, especially at high pH.
GE Healthcare Octyl Sepharose™ 4 Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins
GE Healthcare SOURCE™ 15S Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides
GE Healthcare Capto™ Q Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Designed for capture and intermediate purification of proteins from large feed volumes. GE Heathcare Capto™ Q Chromatography Media is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers.
GE Healthcare Capto™ SP ImpRes BioProcess Media Available on GSA/VA Contract for Federal Government customers only.
A strong cation exchange BioProcess medium for efficient and cost-effective protein purification.
GE Healthcare Capto™ S Chromatography Medium PROMO
Designed for capture and intermediate purification of proteins from large feed volumes. GE Healthcare Capto™ S Chromatography Medium is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong sulfonate (S) cation exchanger
GE Healthcare HiTrap™ TALON™ Crude Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
For purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC).   GE Healthcare Lifescience™ HiTrap™ TALON™ Crude Prepacked Columns offer a cobalt-based medium allowing protein purification under native or denaturing conditions.
  spinner