Chromic Acid
Chromic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Chromic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 1333-82-0 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Liquid
  spinner