Chromium -Cr
Chromium(III) chloride, 99%, sublimed, anhydrous, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium(III) chloride Name Note: 99% CAS: 10025-73-7 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: Cl3Cr Linear Formula: CrCl3 Formula Weight: 158.36 MDL Number: MFCD00010948 Synonym: Chromic chloride
Chromium nitride
Chemical Name or Material: Chromium nitride CAS: 12053-27-9 Molecular Formula: Mixture of Cr2N and CrN MDL Number: MFCD00016048
Chromium(III) nitrate nonahydrate, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium(III) nitrate nonahydrate Name Note: 99% CAS: 2150954 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: CrN3O9·9H2O Linear Formula: Cr(NO3)3·9H2O Formula Weight: 400.14 MDL Number: MFCD00149659 Synonym: Chromic nitrate
Reinecke Salt, ACS Reagent, ACROS Organics™
Chemical Name or Material: Reinecke salt CAS: 13573-16-5 Purity Grade: ACS Reagent Grade Molecular Formula: C4H10CrN7S4·H2O Linear Formula: NH4(Cr(NH3)2(SCN)4)·H2O Formula Weight: 354.46 MDL Number: MFCD00150925 Synonym: Ammonium reineckate
Chromium(III) chloride hexahydrate, 98%, ACROS Organics™
Chemical Name or Material: Chromium(III) chloride hexahydrate Assay: ≥97.5% (CrCl3.6H2O) Assay Percent Range: 98% Molecular Formula: Cl3Cr·6H2O Linear Formula: [Cr(H2O)4Cl2]Cl·2H2O Formula Weight: 266.44 MDL Number: MFCD00149660 Synonym: Chromic chloride hexahydrate
Chromium Trioxide (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium Trioxide CAS: 1333-82-0 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥98.0% Assay Percent Range: ≥98.0 % Molecular Formula: CrO3 Formula Weight: 99.99 MDL Number: MFCD00010952 Synonym: Chromic Acid Anhydride
Chromium(VI) oxide, 99.5%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium(VI) oxide Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: CrO3 Linear Formula: CrO3 Formula Weight: 99.99 MDL Number: MFCD00010952 Synonym: Chromic anhydride
Chromium(III) sulfate hydrate, Dark purple crystals, Alfa Aesar™
Chemical Name or Material: Chromium(III) sulfate hydrate CAS: 15244-38-9 Purity Grade: Reagent Grade Physical Form: Crystals Molecular Formula: Cr2(SO4)3·xH2O Formula Weight: 392.17 (Anhydrous) MDL Number: MFCD00016039
Chromium(III) chloride, Violet flakes, 99.9% (Metals basis), Alfa Aesar™
Chemical Name or Material: Chromium(III) chloride CAS: 10025-73-7 Physical Form: Flakes Assay Percent Range: 99.9% (Metals basis) Molecular Formula: CrCl3 Formula Weight: 158.35 MDL Number: MFCD00010948
Chromium potassium sulfate dodecahydrate, 99+%, ACROS Organics™
Chemical Name or Material: Chromium potassium sulfate dodecahydrate Name Note: p.a. CAS: 7788-99-0 Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CrKO8S2·12H2O Linear Formula: CrK(SO4)2·12H2O Formula Weight: 499.39 MDL Number: MFCD00149917 Synonym: Chrome alum
Chromium etchant, Alfa Aesar™
Chemical Name or Material: Chromium etchant Physical Form: Liquid MDL Number: MFCD07370678
Chromium pieces, electrolytic, 6mm & down, Puratronic™, 99.997% (metals basis)
Chemical Name or Material: Chromium pieces CAS: 7440-47-3 Name Note: Electrolytic Purity Grade: Puratronic™ MDL Number: MFCD00010944
Chromium(VI) oxide, ACS, 98+%
Chemical Name or Material: Chromium oxide CAS: 1333-82-0 Name Note: For analysis ACS Purity Grade: ACS Grade Molecular Formula: CrO3 Formula Weight: 99.99 MDL Number: MFCD00010952 UN Number: UN1463 Merck Index: 14,2235 Synonym: Chromic acid
Chromium Nitrate, Nonahydrate Products for Science Education
Chemical Name or Material: Chromium Nitrate, Nonahydrate CAS: 7789-02-8 Purity Grade: Reagent Grade Linear Formula: Cr(NO3)3·9H2O Formula Weight: 400.18 Synonym: Chromic Nitrate Nonahydrate
Chromium(III) chloride tetrahydrofuran complex (1:3) 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium(III) chloride tetrahydrofuran complex (1:3) Assay Percent Range: 98% Molecular Formula: C12H24Cl3CrO3 Formula Weight: 374.67 MDL Number: MFCD00013222
Chromium powder, 99.99% (metals basis), Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 Physical Form: Powder Assay Percent Range: 99.996% (Metals basis) MDL Number: MFCD00010944
Chromium Powder, -100 mesh, 99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 Physical Form: Powder Assay Percent Range: 99% (Metals basis) MDL Number: MFCD00010944
Chromium(III) oxide, 99.6% (metals basis), Alfa Aesar™
Chemical Name or Material: Chromium oxide CAS: 1308-38-9 Physical Form: 3 to 6mm Fused Lump Assay Percent Range: 99.6% (metals basis) Molecular Formula: Cr2O3 Formula Weight: 151.99 MDL Number: MFCD00010949
Chromium(II) chloride, 90%, ACROS Organics™
Chemical Name or Material: Chromium(II) chloride Name Note: 90% CAS: 10049-05-5 Assay: ≥88% Assay Percent Range: 90% Molecular Formula: Cl2Cr Linear Formula: CrCl2 Formula Weight: 122.9 MDL Number: MFCD00010947 Synonym: Chromous chloride
Chromium Powder, -200 mesh, 99.95% (Metals basis), Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 Physical Form: Powder Assay Percent Range: 99.95% (Metals basis) MDL Number: MFCD00010944
Chromium silicide, >99% (metals basis), Alfa Aesar™
Chemical Name or Material: Chromium silicide CAS: 12018-09-6 Physical Form: Powder Assay Percent Range: >95% Molecular Formula: CrSi2 Formula Weight: 108.17 MDL Number: MFCD00168070
Chromium Powder, -60 mesh, 99.99% (Metals basis), Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 Physical Form: Powder Assay Percent Range: 99.99% (Metals basis) MDL Number: MFCD00010944
Chromium(II) chloride, 99.9%, (trace metal basis), ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chromium(II) chloride CAS: 10049-05-5 Purity Grade: Trace Metal Basis Assay Percent Range: 99.9% Molecular Formula: Cl2Cr Linear Formula: CrCl2 Formula Weight: 122.9 MDL Number: MFCD00010947 Synonym: Chromous chloride
Chromium pieces, 99% (metals basis), Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 Physical Form: Irregular Pieces Assay Percent Range: 99% (Metals basis) MDL Number: MFCD00010944
Chromium(III) acetate hydroxide, Blue crystals, Alfa Aesar™
Chemical Name or Material: Chromium(III) acetate hydroxide Name Note: Cr 24% CAS: 39430-51-8 Physical Form: Crystals Molecular Formula: Cr3(OH)2(OOCCH3)7 Formula Weight: 603.31 MDL Number: MFCD00058912
Chromium (III) Oxide Products for Science Education
Chemical Name or Material: Chromium (III) Oxide CAS: 1308-38-9 Assay Percent Range: 99% w/v Linear Formula: Cr2O3 Formula Weight: 151.99 Synonym: Chromic Oxide
Chromium(III) phosphate tetrahydrate, Alfa Aesar™
Chemical Name or Material: Chromium(III) phosphate tetrahydrate CAS: 10101-59-4 Physical Form: Powder Molecular Formula: CrPO4·4H2O Formula Weight: 219.04 (146.97 Anhydrous) MDL Number: MFCD00016049
Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Alfa Aesar™
Chemical Name or Material: Chromium(III) potassium sulfate dodecahydrate CAS: 7788-99-0 Physical Form: Crystalline Assay Percent Range: 99.9% (Metal basis) Molecular Formula: CrKO8S2·12H2O Formula Weight: 499.39 (283.21 Anhydrous) MDL Number: MFCD00149917
Alfa Aesar™ Chromium powder, 99.99% (metals basis), -100+325 mesh, Alfa Aesar™
Chemical Name or Material: Chromium CAS: 7440-47-3 MDL Number: MFCD00010944
Chromium Products for Science Education
Chemical Name or Material: Chromium CAS: 7440-47-3 Linear Formula: Cr Formula Weight: 52 Synonym: Chrome
  spinner