Citric Acid
Citric acid, disodium salt sesquihydrate, 99%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, disodium salt sesquihydrate Name Note: Pure CAS: 1545832 Purity Grade: Extra Pure Assay: 98.5 to 104.0% (Total base) Assay Percent Range: 99% Molecular Formula: 1·5 H2O Linear Formula: NaO2CCH2C(OH)(CO2H)CH2CO2Na·1·5H2O Formula Weight: 263.1 MDL Number: MFCD00150445 Synonym: Disodium hydrogen citrate sesquihydrate
Citric acid, for molecular biology, anhydrous, Rnase and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid CAS: 77-92-9 Purity Grade: For molecular biology Assay: ≥99% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13 MDL Number: MFCD00011669 Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid Monohydrate (Granular/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: 99.0 to 102.0% Assay Percent Range: 99.0 to 102.0 % Molecular Formula: C6H8O7 · H2O Linear Formula: (HO2CCH2)2C(OH)(CO2H)·H2O Formula Weight: 210.14 MDL Number: MFCD00149972
Citric acid, trisodium salt dihydrate, 99%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt dihydrate CAS: 6132-04-3 Purity Grade: Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C6H5Na3O7·2H2O Linear Formula: NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O Formula Weight: 294.09 MDL Number: MFCD00150031 Synonym: Sodium citrate tribasic dihydrate
Citric acid, anhydrous, ACS, >99.5%, Alfa Aesar™
Chemical Name or Material: Citric Acid Name Note: For analysis ACS CAS: 77-92-9 Physical Form: Crystalline Assay Percent Range: >99.5+% Molecular Formula: C6H8O7 Formula Weight: 192.13 MDL Number: MFCD00011669
Citric acid, triammonium salt, +97%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, triammonium salt CAS: 3458-72-8 Purity Grade: Extra Pure Assay: ≥97.0% Assay Percent Range: 97+% Molecular Formula: C6H17N3O7 Formula Weight: 243.22 MDL Number: MFCD00036406 Synonym: Triammoniumcitrate
Citric Acid, Anhydrous p.a., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid CAS: 77-92-9 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13 Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid, Anhydrous, Reagent ACS 99.5%, ACROS Organics™
Chemical Name or Material: Citric acid CAS: 77-92-9 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13 Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid Anhydrous (Crystalline/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid, Anhydrous CAS: 77-92-9 Purity Grade: USP Physical Form: Crystalline Assay: 99.5 to 100.5% (USP) Molecular Formula: C6H8O7 Formula Weight: 192.13 MDL Number: MFCD00011669
Citric Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 Purity Grade: Certified ACS Grade Physical Form: Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13 MDL Number: MFCD00011669
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt hydrate Name Note: For molecular biology CAS: 114456-61-0 Purity Grade: For molecular biology Assay: ≥99.75% (on dry substance) Assay Percent Range: 99.8% Molecular Formula: C6H5Na3O7·xH2O Formula Weight: 258.08 MDL Number: MFCD00150032
Citric acid monohydrate, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: Citric acid monohydrate CAS: 5949-29-1 Purity Grade: For analysis Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: C6H8O7·H2O Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H·H2O Formula Weight: 210.15 MDL Number: MFCD00149972
Citric Acid, Anhydrous Products for Science Education
Chemical Name or Material: Citric Acid, Anhydrous CAS: 77-92-9 Purity Grade: Laboratory Grade Assay Percent Range: 1% Linear Formula: C6H8O7 Formula Weight: 192.13 Synonym: β-Hydroxytricarboxylic Acid
Citric acid, trisodium salt dihydrate, 99%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt dihydrate CAS: 6132-04-3 Purity Grade: For biochemistry Assay Percent Range: 99% Molecular Formula: C6H5Na3O7·2H2O Linear Formula: NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O Formula Weight: 294.09 Synonym: Sodium citrate tribasic dihydrate
Citric acid, diammonium salt, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, diammonium salt CAS: 3012-65-5 Purity Grade: Extra Pure Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H14N2O7 Formula Weight: 226.19 MDL Number: MFCD00013068 Synonym: Diammoniumcitrate
Citric Acid Monohydrate (Granular/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: USP Physical Form: Solid Assay: 99.5 to 100.5% (USP) Molecular Formula: C6H8O7 · H2O Formula Weight: 210.14 MDL Number: MFCD00149972
Citric Acid, Anhydrous, 99%, Pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid Name Note: Anhydrous CAS: 77-92-9 Purity Grade: Pure Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C6H8O7 Linear Formula: HO2CCH2C(OH)(CO2H)CH2CO2H Formula Weight: 192.13 MDL Number: MFCD00011669 Synonym: 2-Hydroxy-1, 2, 3-propanetricarboxylic acid
Citric Acid Monohydrate (Crystalline/USP/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Monohydrate CAS: 5949-29-1 Purity Grade: BP/EP/JP/USP Physical Form: Solid Molecular Formula: C6H8O7 · H2O Formula Weight: 210.14 MDL Number: MFCD00008765
Citric acid, trisodium salt 98%, anhydrous, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric acid, trisodium salt CAS: 68-04-2 Purity Grade: Pure Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C6H5Na3O7 Formula Weight: 258.07 MDL Number: MFCD00012462 Synonym: Trisodium citrate
Citric Acid , Monohydrate Products for Science Education
Chemical Name or Material: Citric Acid, Monohydrate CAS: 5949-29-1 Assay Percent Range: 1% Linear Formula: C6H8O7·H2O Formula Weight: 210.15 Synonym: 2-Hydroxy-1, 2, 3-Propanetricarboxylic Acid Monohydrate
Citric Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 77-92-9 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Liquid
Citric Acid, 1.00 Molar, Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 86.0 CAS: 77-92-9 CAS Min %: 15.8 CAS Max %: 16.45 Physical Form: Liquid
Citric acid, Tocris
Chemical Name or Material: Citric acid CAS: 77-92-9 Assay Percent Range: >99.5% Molecular Formula: C6H8O7 Formula Weight: 192.12 Synonym: 2-Hydroxy-1, 2, 3-propanenetricarboxylic acid
Citric Acid U.S.P. Granular, Macron Fine Chemicals™
Chemical Name or Material: Citric Acid Name Note: Monohydrate CAS: 5949-29-1 Purity Grade: USP Physical Form: Granular Molecular Formula: C6H8O7·H2O Formula Weight: 210.14
Citric Acid Anhydrous ACS, Ricca Chemical
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS Grade Physical Form: Granules
Citric Acid Anhydrous (Crystalline), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Citric Acid Anhydrous CAS: 77-92-9 Physical Form: Solid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C6H8O7 Formula Weight: 192.13
Citric Acid, 1% (w/w), Ricca Chemical
Chemical Name or Material: Citric Acid CAS: 77-92-9 CAS: 7732-18-5
Citric Acid, Monohydrate Granular/ACS), Macron Fine Chemicals™
Chemical Name or Material: Citric Acid, Monohydrate CAS: 5949-29-1 Purity Grade: ACS Grade Physical Form: Granular Molecular Formula: C6H8O7·H2O Formula Weight: 210.14
Citric acid, 99.91%, MP Biomedicals™
Chemical Name or Material: Citric acid CAS: 5949-29-1 Physical Form: Crystalline Powder Molecular Formula: C6H8O7·H2O Formula Weight: 210.1 Synonym: β-hydroxytricarballylic acid
Sodium citrate, 99.8%, For ACS analysis, MP Biomedicals™
Chemical Name or Material: Sodium citrate Name Note: For ACS analysis CAS: 6132-04-3 Physical Form: Crystalline Powder Molecular Formula: C6H5Na3O7.2H2O Formula Weight: 294.1 Synonym: Citric acid, trisodium salt, dihydrate
  spinner