Concentrated Buffers
Thermo Scientific™ Pierce™ 20X TBS Tween™-20
Prepare TBS-Tween (TBS-T) wash buffers or blocking buffers for ELISA, Western blotting and other immunoassays with this space-saving stock solution.
Thermo Scientific™ Pierce™ 10X Tris-Glycine-SDS Buffer
Quickly prepare Tris-glycine-SDS (pH 8.3) running buffer for SDS-PAGE (protein polyacrylamide gel electrophoresis) with this space-saving stock solution.
Thermo Scientific™ Pierce™ 10X TBE Buffer
Cast agarose gels or prepare Tris-borate-EDTA (pH 8.3) running buffer for agarose and polyacrylamide gel electrophoresis with this space-saving stock solution.
Thermo Scientific™ Pierce™ 10X Tris-Glycine Buffer
Quickly prepare standard Tris-glycine (pH 8.5) transfer buffer for Western blot membrane with this space-saving stock solution.
Thermo Scientific™ Pierce™ 10X Methanol-free Western Blot Transfer Buffer
A methanol-free concentrated solution to easily and quickly prepare MeOH-free Western blot transfer buffer for tank-based or other wet gel-to-membrane transfer methods.
Thermo Scientific™ Pierce™ 20X Modified Dulbecco's PBS Tween-20
Prepare Na- and K-phosphate buffered saline (D-PBS) wash and blocking buffers for ELISA, Western and other immunoassays with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X Tris-HEPES-SDS Buffer
Prepare Tris-HEPES-SDS running buffer required for gel electrophoresis (SDS-PAGE) with Precise Precast Gels with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X Tris Buffered Saline
Prepare Tris-NaCl buffer (TBS) used as a wash buffer and antibody diluent for ELISA, Western blotting and other immunoassays with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X TAE Buffer
Cast agarose gels and prepare Tris-Acetate EDTA (pH 8.3) running buffer for nucleic acid electrophoresis with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X Modified Dulbecco's PBS
Prepare Na- and K-phosphate buffered saline (D-PBS) diluents for ELISA, Western blotting and other immunoassays with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X Phosphate Buffered Saline
Prepare PBS buffers for crosslinking, biotinylation and fluorescent labeling reactions requiring an amine-free buffer with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X PBS Tween-20
Prepare PBS-Tween (PBS-T) wash buffers and blocking buffers for ELISA and Western with this space-saving stock solution.
Thermo Scientific™ Pierce™ 20X Borate Buffer
Prepare sodium borate buffer solutions for protein modification procedures requiring amine-free buffer at alkaline pH with this space-saving stock solution.
R&D Systems ExactaChIP™ Chromatin IP Buffer Panel
For chromatin immunoprecipitation using the complete kit
R&D Systems Ubiquitin Conjugation Reaction Buffer Kit
Contains the optimal buffer formulations for use in conjugation assays of ubiquitin to protein substrates in vitro
R&D Systems NEDD8 Conjugation Reaction Buffer Kit
Contains the optimal buffer formulations for use in assays for the conjugation of the ubiquitin-like modifier NEDD8 to protein substrates in vitro
R&D Systems SUMO Conjugation Reaction Buffer Kit
Contains the optimal buffer formulations for use in assays for the conjugation of the ubiquitin-like modifiers, SUMO-1, SUMO-2, and SUMO-3 to protein substrates in vitro
Lonza ProSieve™ EX Transfer Buffer for Western Blot
Complete transfer in just 10-15 minutes
Lonza ProSieve™ EX Running Buffer
Novel formulation of the classic Laemmli buffer
  spinner