Conductivity and TDS Standards
Potassium Chloride Conductivity Standard, 50μS/cm at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 50μS/cm at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.0025 CAS Max %: 0.0026 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 500μS/cm (500μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Name Note: Conductivity Standard CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.03 CAS Max %: 0.03 Physical Form: Liquid
Conductivity Standard, 10μS/cm (10μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Conductivity Standard Name Note: 10μS/cm (10μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 5.0E-4 CAS Max %: 5.0E-4 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 50,0000μS/cm (50,000μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 50, 0000μS/cm (50, 000μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 95.0 CAS Max %: 99.0 CAS: 7447-40-7 CAS Min %: 3.02 CAS Max %: 3.14 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 718 μS/cm (718 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 718μS/cm (718μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.04 CAS Max %: 0.04 Physical Form: Liquid
Conductivity/TDS Standard, 500μS/cm (500μmho/cm) at 25°C, 240ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard Name Note: 500μS/cm (500μmho/cm) at 25°C Physical Form: Liquid
Conductivity/TDS Standard, 100μS/cm at 25°C, 47.2ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard Name Note: 100μS/cm (100μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.0046 CAS Max %: 0.0048 Physical Form: Liquid
Conductivity Standard, 10 mS/cm (10,000 μmho/cm) at 25°C, with attached Certificate of Analysis, Ricca Chemical
Chemical Name or Material: Conductivity Standard Name Note: With attached Certificate of Analysis CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.53 CAS Max %: 0.55 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 2000 μS/cm (2,000 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 2000μS/cm (2, 000μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.1 CAS Max %: 0.11 Physical Form: Liquid
Conductivity/TDS Standard, 10mS/cm (10,000μmho/cm) at 25°C, 5400ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard Name Note: 10mS/cm ((10, 000μmho/cm) at 25°C, 5400ppm as NaCl CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.53 CAS Max %: 0.55 Physical Form: Liquid
Potassium Chloride Conductivity Standard,1,413 μmho/cm at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 1, 413 μmho/cm at 25°C CAS: Potassium Chloride 7447-40-7 CAS Max %: 15.0 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 10 mS/cm (10,000 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 10 mS/cm (10, 000μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.56 CAS Max %: 0.59 Physical Form: Liquid
Potassium Chloride Reference Solution C, 1408.8 μS/cm (1408.8 μmho/cm) at 25°C, 0.01 Molar, Ricca Chemical
Chemical Name or Material: Potassium Chloride Reference Solution C Name Note: 1408.8μS/cm (1408.8μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.07 CAS Max %: 0.07 Physical Form: Liquid
Conductivity/TDS Standard, 16.63 mS/cm (16,630 μmho/cm) at 25°C, 9100ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.92 CAS Max %: 0.96 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 1 mS/cm (1000 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 1 mS/cm (1000μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.05 CAS Max %: 0.05 Physical Form: Liquid
Conductivity/TDS Standard, 3 mS/cm (3000 μmho/cm) at 25°C, 1534ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.15 CAS Max %: 0.16 Physical Form: Liquid
Conductivity/TDS Standard, 50 mS/cm (50,000 μmho/cm) at 25°C, 30,300ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard CAS: 7647-14-5 (Sodium Chloride) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Conductivity/TDS Standard, 150 μS/cm (150 μmho/cm) at 25°C, 71ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard CAS: 7647-14-5 (Sodium Chloride) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Potassium Chloride Conductivity Standard, 25,000μS/cm (25,000μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 25, 000μS/cm (25, 000μmho/cm) at 25°C CAS: 7732-18-5 CAS: 7447-40-7 Physical Form: Liquid Assay Percent Range: 25, 000 ± 100 μS/cm at 25°C
Potassium Chloride Conductivity Standard, 200 μS/cm (200 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 200 μS/cm (200 μmho/cm) at 25°C CAS: 7447-40-7 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 200 ± 1 μS/cm at 25°C
Potassium Chloride Conductivity Standard, 5 mS/cm (5000 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 5 mS/cm (5000μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.27 CAS Max %: 0.28 Physical Form: Liquid
Conductivity Standard, 1,800 μmhos/cm at 25°C, Ricca Chemical
Chemical Name or Material: Conductivity Standard Name Note: 1, 800μmhos/cm at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7647-14-5 CAS Min %: 0.08 CAS Max %: 0.09 Physical Form: Liquid
Conductivity/TDS Standard, 20 mS/cm (20,000 μmho/cm) at 25°C, 11,000ppm as NaCl, Ricca Chemical
Chemical Name or Material: Conductivity/TDS Standard CAS: 7647-14-5 (Sodium Chloride) CAS: 7732-18-5 (Water, Deionized) Physical Form: Liquid
Potassium Chloride Conductivity Standard, 1.5 mS/cm (1,500μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 1.5 mS/cm (1, 500μmho/cm) at 25°C CAS: 7732-18-5 CAS: 7447-40-7 Physical Form: Liquid Assay Percent Range: 1, 500 ± 1 μS/cm at 25°C
Potassium Chloride Conductivity Standard, 70 mS/cm at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 70 mS/cm at 25°C CAS: 7447-40-7 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 70, 000 ± 100 μS/cm at 25°C
Potassium Chloride Reference Solution D, 146.93 μS/cm (146.93 μmho/cm) at 25°C, 0.001 Molar, Ricca Chemical
Chemical Name or Material: Potassium Chloride Reference Solution D Name Note: 146.93μS/cm (146.93μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.01 CAS Max %: 0.01 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 84 μS/cm (84 μmho/cm) at 25°C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard Name Note: 84μS/cm (84μmho/cm) at 25°C CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.0040 CAS Max %: 0.0042 Physical Form: Liquid
Potassium Chloride Conductivity Standard, 1015 μS/cm (1015 μmho/cm) at 25 °C, Ricca Chemical
Chemical Name or Material: Potassium Chloride Conductivity Standard CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7447-40-7 CAS Min %: 0.05 CAS Max %: 0.05 Physical Form: Liquid
  spinner