Contract Laboratory Program -CLP Standards
Contract Required Detection Limits (CRDL), Ricca Chemical
Chemical Name or Material: Contract Required Detection Limits (CRDL) CAS: 7697-37-2 (Nitric Acid) CAS: 7732-18-5 (Water, Deionized)
Spiking Solution for Water and Soil - CLP, Ricca Chemical
Chemical Name or Material: Spiking Solution CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 95.0 CAS: 7697-37-2 CAS Min %: 6.53 CAS Max %: 6.8 CAS: 7784-27-2 CAS Min %: 0.18 CAS Max %: 0.19 CAS: 10022-31-8 CAS Min %: 0.03 CAS Max %: 0.03 CAS: 7782-49-2 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 7440-28-0 CAS Min %: 0.01 CAS Max %: 0.01 Purity Grade: CLP Physical Form: Liquid
Primary Analytes Standard, CLP, Ricca Chemical
Chemical Name or Material: Primary Analytes Standard CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 95.0 CAS: 7697-37-2 CAS Min %: 6.77 CAS Max %: 7.05 CAS: 7789-02-8 CAS Min %: 0.03 CAS Max %: 0.03 CAS: 7440-43-9 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 7440-66-6 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 10022-31-8 CAS Min %: 0.01 CAS Max %: 0.01 Purity Grade: CLP Physical Form: Liquid
Spike Sample Analysis Standard I For CLP, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ICP Spike Sample 1 For Use With (Equipment): AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
UOP Method 551 Standard-7 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 551 Standard
UOP Method 690 Standard-7 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 690 Standard
Semivolatiles Balance Mix- Option A, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Semi-Volatile Balance Mix - Option A
Volatiles Supplementary Volatiles Mix, SPEX CertiPrep™
Chemical Name or Material: Supplementary Volatiles Mix
ASTM UOP Method 720 Standard-5 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 720 Standard
ASTM Method D2887 Standard-20 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ASTM Method D2887 Standard
ASTM Method D6296 Standard-11 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ASTM Method D6296 Standard
Alternate Analytes B For CLP, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Alternate Interferents B Purity Grade: For CLP (Contract Laboratory Program)
Instrument Calibration Standard 6 For CLP, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Mercury Standard CAS: 7439-97-6 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 7697-37-2 CAS Min %: 5.0 CAS Max %: 5.0 CAS: 7732-18-5 CAS Min %: 94.99 CAS Max %: 94.99 Purity Grade: For CLP (Contract Laboratory Program)
Volatiles Organics Combination Standard, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Volatile Organics Combination Standard
Initial Calibration Verification Standard IA For CLP, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Initial Calibration Verification Standard 1A Purity Grade: For CLP (Contract Laboratory Program)
UOP Method 744 Standard-8 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 744 Standard
Volatiles Mix A, SPEX CertiPrep™
Chemical Name or Material: Volatile Analyte Mix A
CLP 90 Mix F, SPEX CertiPrep™
Chemical Name or Material: CLP 90 Mix F
Vinyl Acetate, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Vinyl acetate Name Note: Methods 8240B, 8260B, CLP Methods CAS: 108-05-4 CAS Min %: 0.1 CAS Max %: 0.1 CAS: 67-56-1 CAS Min %: 99.9 CAS Max %: 99.9
Instrument Calibration Standard 2 For CLP, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Instrument Calibration Standard 2 Purity Grade: For CLP (Contract Laboratory Program)
UOP Method 931 Standard-5 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 931 Standard
Combination Semi-Volatile Surrogate Standard, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Combination Semivolatile Surrogate Standard Name Note: CLP SOW 2/88
ASTM Method D3710 Standard-16 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: ASTM Method D3710 Standard
Semi-Volatile Calibration Standard, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Semi-volatile Calibration Standard For Use With (Equipment): GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Purgeable Gases, Mix B, High Level, SPEX CertiPrep™
Chemical Name or Material: Purgeable Gases Mix B (High Level) Name Note: Method 8240B, CLP Methods CAS: Various
Alternate ASTM Method D6296 Standard-10 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Alternate ASTM Method D6296 Standard
91-SOW Surrogate Spike, SPEX CertiPrep™
Chemical Name or Material: 91 SOW Surrogate Spike Name Note: CLP Methods
Surrogate Standard, High Level, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Surrogate Standard (High Level) Purity Grade: For CLP series volatiles
UOP Method 543 Standard-2 Components, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: UOP Method 543 Standard
High Concentration Base/Neutrals Surrogate, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: High Conc Base/Neutrals Surrogate Standard
  spinner