Cyanuric Acid
Cyanuric acid, 98%, ACROS Organics™
Chemical Name or Material: Cyanuric acid Name Note: 98% CAS: 108-80-5 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C3H3N3O3 Formula Weight: 129.07 MDL Number: MFCD00082990 Synonym: 1,3,5-Triazine-2,4,6-triol
Cyanuric Acid, 99%, Alfa Aesar™
Chemical Name or Material: Cyanuric Acid Assay Percent Range: 99% Molecular Formula: C3H3N3O3 Formula Weight: 129.08
  spinner