Cyclohexane
Cyclohexane, 99+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane (Reagent), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Reagent Grade Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, 99.0%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.0% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, >99%, ACS, Alfa Aesar™
Chemical Name or Material: Cyclohexane Name Note: For ACS analysis CAS: 110-82-7 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C6H12 Formula Weight: 84.16 MDL Number: MFCD00003814
Cyclohexane, 99.8%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: For HPLC Assay: ≥99.8% (GC) Assay Percent Range: 99.8% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, ACROS Organics™
Chemical Name or Material: Cyclohexane Name Note: ECD Tested, For Pesticide Analysis CAS: 110-82-7 Purity Grade: For residue analysis Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexene Products for Science Education
Chemical Name or Material: Cyclohexene CAS: 110-83-8 Assay Percent Range: 0.9999 Linear Formula: C6H12 Formula Weight: 82.13
Cyclohexane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane Name Note: Extra Dry CAS: 110-82-7 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, 99+%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: ACS Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane Products for Science Education
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Reagent Grade Assay Percent Range: >95% w/v Linear Formula: C6H12 Formula Weight: 84.15 Synonym: Hexahydrobenzene
Cyclohexane (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Certified ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: ≥99.0% Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane (HPLC), Fisher Chemical
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.0% Assay Percent Range: 99.0% min by GC Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, HPLC Grade, 99% min.; Alfa Aesar™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: C6H12 Formula Weight: 84.16 MDL Number: MFCD00003814
Cyclohexane, extra pure, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Pure Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15
Cyclohexane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane Name Note: 99+% CAS: 110-82-7 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane (Pesticide), Fisher Chemical
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Pesticide Grade Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, for residue analysis, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: For residue analysis Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, for HPLC, ACROS Organics™
Chemical Name or Material: Cyclohexane Name Note: 99.0% CAS: 110-82-7 Purity Grade: For HPLC Assay: ≥99.0% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane, AR ACS), For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Cyclohexane Name Note: ACS CAS: 110-82-7 Purity Grade: ACS Reagent Grade Assay Percent Range: 99.0 % min. Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane Name Note: For ACS analysis CAS: 110-82-7 Purity Grade: ACS/NF/USP Physical Form: Liquid Assay Percent Range: 99.0%, min. Molecular Formula: C6H12 Linear Formula: -CH2(CH2)4CH2- Formula Weight: 84.16 Synonym: Hexahydrobenzene
Cyclohexane, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.18 UN Number: UN 1145 Synonym: Hexahydrobenzene
Cyclohexane Nanograde, Macron™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane ACS AR, Macron™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: ACS Reagent Grade Molecular Formula: C6H12 Formula Weight: 84.16
  spinner