Cyclohexanone
Cyclohexanone, ≥99%, Alfa Aesar™
Chemical Name or Material: Cyclohexanone CAS: 108-94-1 Physical Form: Liquid Assay Percent Range: ≥99% Molecular Formula: C6H10O Formula Weight: 98.15
Cyclohexanone, 99.8%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexanone CAS: 108-94-1 Purity Grade: Extra Pure Assay: ≥99.75% Assay Percent Range: 99.8% Molecular Formula: C6H10O Linear Formula: C6H10(=O) Formula Weight: 98.14 MDL Number: MFCD00001625
Cyclohexanone Oxime 97%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexanone oxime Name Note: 97% CAS: 100-64-1 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C6H11NO Linear Formula: C6H10(=NOH) Formula Weight: 113.16 MDL Number: MFCD00001660
Cyclohexanone (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexanone CAS: 108-94-1 Purity Grade: Certified Grade Assay: ≥98% Assay Percent Range: ≥98 % Molecular Formula: C6H10O Formula Weight: 98.14 MDL Number: MFCD00001625 Synonym: Ketohexamethylene
Cyclohexanone (Laboratory), Fisher Chemical
Chemical Name or Material: Cyclohexanone CAS: 108-94-1 Purity Grade: Laboratory Grade Molecular Formula: C6H10O Linear Formula: CH2(CH2)4CO Formula Weight: 98.14 MDL Number: MFCD00001625 Synonym: Ketohexamethylene
Cyclohexanone ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Cyclohexanone CAS: 108-94-1 Purity Grade: ACS Reagent Grade Molecular Formula: C6H10O Formula Weight: 98.14
Cyclohexanone, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexanone Name Note: For ACS analysis CAS: 108-94-1 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: -CH2(CH2)4C(:O)- Linear Formula: -CH2(CH2)4C(:O)- Formula Weight: 98.14 Synonym: Pimelic Ketone
  spinner