Cysteine -Cys and Derivatives
D-Cysteine, 99%, ACROS Organics™
Chemical Name or Material: D-Cysteine CAS: 921-01-7 Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C3H7NO2S Linear Formula: HSCH2CH(NH2)CO2H Formula Weight: 121.16 MDL Number: MFCD00066461
L-Cysteine Hydrochloride Hydrate, 98.5-101.5%, ACROS Organics™
Chemical Name or Material: L-Cysteine hydrochloride hydrate Assay: 98.5 to 101.0% (on dried substance) Assay Percent Range: 98.5-101.0% Molecular Formula: C3 H7 N O2 S . H Cl . x H2 O Linear Formula: HCl·xH2O Formula Weight: 157.61 MDL Number: MFCD00065606
Se-methylseleno-L-cysteine, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Se-methylseleno-L-cysteine CAS: 26046-90-2 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H9NO2Se Formula Weight: 182.08 MDL Number: MFCD00800565
BOC-S-trityl-L-cysteine, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: BOC-S-trityl-L-cysteine Assay Percent Range: 99% Molecular Formula: C27H29NO4S Formula Weight: 463.6 MDL Number: MFCD00038251
L-Cysteine, 98+%
Chemical Name or Material: L-Cysteine CAS: 52-90-4 Molecular Formula: C3H7NO2S Formula Weight: 121.16 MDL Number: MFCD00064306 Beilstein: 1721408 Merck Index: 14,2781
DL-Cysteine 99%, ACROS Organics™
Chemical Name or Material: DL-Cysteine Name Note: 99% CAS: 3374-22-9 Assay Percent Range: 97% Molecular Formula: C3H7NO2S Linear Formula: HSCH2CH(NH2)CO2H Formula Weight: 121.15 Synonym: 2-Amino-3-mercaptopropionic acid
D-Cysteine hydrochloride monohydrate, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 207121-46-8 Physical Form: Crystalline Powder Assay Percent Notes: 98% Molecular Formula: H20·HCl Formula Weight: 177.77 MDL Number: MFCD00150051
S-Sulfo-L-cysteine sodium salt, Tocris
Chemical Name or Material: S-Sulfo-L-cysteine sodium salt CAS: 7381-67-1 Assay Percent Range: >98% Molecular Formula: C3H6NO5S2Na Formula Weight: 223.19
L-Cysteine Hydrochloride Monohydrate (White Crystals), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-Cysteine Hydrochloride Monohydrate CAS: 7048-04-6 Physical Form: Solid Assay: 98.5 to 101% Assay Percent Range: 98.5 to 101 % Molecular Formula: C3H7NO2S · HCl · H2O Formula Weight: 175.64
Lactacystin, Tocris
Chemical Name or Material: Lactacystin CAS: 133343-34-7 Assay Percent Range: >98% Molecular Formula: C15H24N2O7S Formula Weight: 376.42 Synonym: (2R, 3S, 4R)-3-Hydroxy-2-[(1S)-1-hydroxy-2-methylpropyl]-4-methyl-5-oxo-2-pyrrolidinecarboxy-N-acetyl-L-cysteine thioester
N-acetyl-L-cysteine, >96%, MP Biomedicals™
Chemical Name or Material: N-acetyl-L-cysteine CAS: 616-91-1 Physical Form: Crystalline Powder Assay Percent Range: >96% Molecular Formula: C5H9NO3S Formula Weight: 163.2 MDL Number: MFCD00004880 Synonym: 1alpha;-Acetamido-β-mercaptopropionic acid
L-Cysteine, 99.5%, MP Biomedicals™
Chemical Name or Material: L-Cysteine CAS: 52-90-4 Physical Form: Crystalline Powder Assay Percent Range: 99.5% Molecular Formula: C3H7NO2S Formula Weight: 121.16 Synonym: β-Mercapto-L -alanine
N-Acetyl-L-cysteine (Crystalline), MP Biomedicals
Chemical Name or Material: N-Acetyl-L-cysteine CAS: 616-91-1 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Powder Assay Percent Notes: ≥96% Molecular Formula: C5H9NO3S Formula Weight: 163.2 MDL Number: MFCD00004880 Synonym: 1-alpha-Acetamido-beta-mercaptopropionic acid
L-Cysteine hydrochloride monohydrate, MP Biomedicals™
Chemical Name or Material: L-Cysteine Hydrochloride Monohydrate Name Note: Precursor in the food, pharmaceutical, personal care industries CAS: 7048-04-6 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystals Molecular Formula: C3H7NO2S · HCl · H2O Formula Weight: 175.64 MDL Number: MFCD00065606 Synonym: Cysteine chlorhydrate
L-Cysteine [Free Base], MP Biomedicals
Chemical Name or Material: L-Cysteine [Free Base] CAS: 52-90-4 Purity Grade: Cell Culture Reagent Grade Physical Form: Crystalline Molecular Formula: C3H7NO2S Formula Weight: 121.2 Synonym: β-Mercapto-L-alanine
DL-Cystine, Free acid, MP Biomedicals™
Chemical Name or Material: DL-Cystine Name Note: Free Acid CAS: 923-32-0 Molecular Formula: C6H12N2O4S2 Formula Weight: 240.3 MDL Number: MFCD00084652 Synonym: (±)-3, 3'-Dithiobis(2-aminopropionic acid)
L-Cysteine, 99+%, ACROS Organics™
Chemical Name or Material: L-Cysteine Assay: ≥99.0% (on dried substance) Assay Percent Range: 98+% Molecular Formula: C3H7NO2S Linear Formula: HSCH2CH(NH2)CO2H Formula Weight: 121.15 MDL Number: MFCD00064306 Synonym: (+)-2-Amino-3-mercaptopropionic acid
FMOC-S-trityl-L-Cysteine, 97%, Acros Organics
Chemical Name or Material: FMOC-S-trityl-L-Cysteine Name Note: 97% CAS: 103213-32-7 Assay: ≥96.0% Assay Percent Range: 97% Molecular Formula: C37H31NO4S Formula Weight: 585.72 MDL Number: MFCD00038538
DL-Cysteine hydrochloride hydrate, 99%, ACROS Organics™
Chemical Name or Material: DL-Cysteine hydrochloride hydrate Name Note: 99% CAS: 96998-61-7 Assay: 98.5 to 101.5% (on dried substance) Assay Percent Range: 99% Molecular Formula: HCl·xH2O Linear Formula: HCl·xH2O MDL Number: MFCD00151030
  spinner