Desiccants and Drying Agents
Silica gel, for chromatography, 0.030-0.200 mm, 60 A, ACROS Organics™
Chemical Name or Material: Silica gel Name Note: 0.060-0.200mm, 60 A CAS: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.08 MDL Number: MFCD00011232 Synonym: Silicon dioxide
Silica Gel Desiccant Indicating(Grade 48, 4-10 Mesh/Certified ACS), Fisher Chemical
Chemical Name or Material: Silica Gel Desiccant, Indicating Name Note: 4-10 Mesh, Grade 48 CAS: 112926-00-8 CAS: 7646-79-9 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: SiO2 MDL Number: MFCD00011232
Alfa Aesar™ Drierite, regular (CaSO4;), -6 Mesh, Lab Grade, Alfa Aesar™
Chemical Name or Material: Drierite CAS: 7778-18-9 Purity Grade: Laboratory Grade Molecular Formula: CaSO4 MDL Number: MFCD00010912
Drierite, with indicator, 8 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drierite™ Name Note: With indicator, 8 Mesh CAS: 7778-18-9 CAS: 7646-79-9 Molecular Formula: CaO4S Formula Weight: 136.13 MDL Number: MFCD00010912
Florisil, 60-100 mesh, for column chromatography, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Florisil™ Name Note: 60-100 mesh, for column chromatography CAS: 1343-88-0 Molecular Formula: MgO3Si Formula Weight: 100.39 MDL Number: MFCD00078249
Silica Gel Desiccant (100-200 Mesh/923, Chromatographic Grade), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Silica Gel Desiccant Name Note: 100-200 Mesh CAS: 7631-86-9 Purity Grade: 923, Chromatographic Grade Molecular Formula: SiO2 MDL Number: MFCD00011232
Alfa Aesar™ Silica Gel Desiccants, -6+12 mesh granules, Alfa Aesar
Chemical Name or Material: Silica gel desiccant CAS: 7631-86-9 MDL Number: MFCD00011232
Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ascarite(II)™, CO2 absorbent, 20-30 mesh, capacity: 40-50% Name Note: 20- to 30-mesh CAS: 81133-20-2 CAS: 1310-73-2 CAS: 7631-86-9 MDL Number: MFCD00132264
Drierite™, with indicator, 10-20 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drierite™ CAS: 7778-18-9 CAS: 7646-79-9 Molecular Formula: CaO4S Formula Weight: 136.13
Alfa Aesar™ Drierite, regular (CaSO4;), -10+20 Mesh, Lab Grade, Alfa Aesar™
Chemical Name or Material: Drierite CAS: 7778-18-9 Purity Grade: Laboratory Grade Molecular Formula: CaSO4 MDL Number: MFCD00010912
Drierite™, Regular (CaSO4), Anhydrous; Lab Grade, Alfa Aesar™
Chemical Name or Material: Calcium Sulfate Name Note: Regular CAS: 7778-18-9 Purity Grade: Laboratory Grade Physical Form: Granular, -10+12 Mesh MDL Number: MFCD00010912 Synonym: Calcium Sulfate, Anhydrous
Silica Gel Desiccant (60-200 Mesh/Chromatographic Grade), Fisher Chemical
Chemical Name or Material: Silica Gel Desiccant Name Note: 60-200 Mesh CAS: 7631-86-9 Purity Grade: Chromatographic Grade Molecular Formula: SiO2 MDL Number: MFCD00011232
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, ACROS Organics™
Chemical Name or Material: Silica gel Name Note: For column chromatography CAS: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.08 Synonym: Silicon dioxide
Silica Gel Desiccant (12-24 mesh, Grade 408/Certified ACS), Fisher Chemical
Chemical Name or Material: Silica Gel Desiccant Name Note: 12-24 Mesh, Grade 408 CAS: 7631-86-9 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: SiO2 MDL Number: MFCD00011232
Calcium Chloride Anhydrous (Pellets, 4-20 mesh, for Desiccators), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Calcium Chloride Anhydrous Name Note: For desiccators CAS: 10043-52-4 Physical Form: Solid Molecular Formula: Ca Cl2 Formula Weight: 110.99 MDL Number: MFCD00010903
Drierite, indicating (CaSO4), Alfa Aesar™
Chemical Name or Material: Drierite CAS: 7778-18-9 Purity Grade: Laboratory Grade Physical Form: Granular Molecular Formula: CaSO4 MDL Number: MFCD00010912
Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Silica gel, functionalized, Amino-3 CAS: 71888-97-6 MDL Number: MFCD03427643
Alfa Aesar™ Silica Gel Indicating Desiccants, -6+16 mesh granules, Alfa Aesar™
Chemical Name or Material: Silica Gel Desiccant CAS: 7631-86-9 Purity Grade: ACS MDL Number: MFCD00011232
Drierite™ Products for Science Education
Chemical Name or Material: Drierite™ CAS: 7778-18-9 Formula Weight: 136.14 Synonym: Calcium Sulfate
Silica Gel Desiccants, Alfa Aesar™
Chemical Name or Material: Silica gel desiccant CAS: 7631-86-9 Physical Form: Granular MDL Number: MFCD00011232
Drierite™, without indicator, 8 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drierite™ CAS: 7778-18-9 Molecular Formula: CaO4S Formula Weight: 136.14
Drierite™, without indicator, 10-20 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drierite™ CAS: 7778-18-9 Molecular Formula: CaO4S Formula Weight: 136.14
Silica Gel Desiccant (6-12 Mesh, Certified ACS), Fisher Chemical
Chemical Name or Material: Silica Gel Desiccant Name Note: 6-12 Mesh CAS: 63231-67-4 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: SiO2 MDL Number: MFCD00011232
Drierite™, without indicator, 20-40 mesh, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drierite™ Name Note: 20-40 mesh CAS: 7778-18-9 Molecular Formula: CaO4S Formula Weight: 136.14 MDL Number: MFCD00010912
Silicagel orange, for drying purposes, non toxic grade, ACROS Organics™
Chemical Name or Material: Silica gel orange CAS: 1327-36-2 MDL Number: MFCD00077645
Drierite, regular (CaSO4), Lab Grade, Alfa Aesar™
Chemical Name or Material: Drierite CAS: 7778-18-9 Purity Grade: Laboratory Grade Physical Form: Granular Molecular Formula: CaSO4 MDL Number: MFCD00010912
Drierite, Blue granules, Alfa Aesar™
Chemical Name or Material: Drierite Name Note: Indicating CAS: 7778-18-9 Physical Form: Granules MDL Number: MFCD00010912
Silica gel, for drying purposes, non-toxic grade, 2-5 mm, ACROS Organics™
Chemical Name or Material: Silica gel Name Note: 2-5mm CAS: 7631-86-9 Purity Grade: Non-toxic Grade Molecular Formula: O2Si Formula Weight: 60.08 MDL Number: MFCD00011232 Synonym: Silicon dioxide
Silica Gel Desiccant (28-200 mesh, Grade 12/Certified ACS), Fisher Chemical
Chemical Name or Material: Silica Gel Desiccant Name Note: 28-200 Mesh, Grade 12 CAS: 63231-67-4 Purity Grade: Certified ACS Grade Physical Form: Solid Molecular Formula: SiO2 MDL Number: MFCD00011232
Silica gel desiccant, indicating, ACS, Alfa Aesar™
Chemical Name or Material: Silica gel desiccant Name Note: For analysis ACS CAS: 7631-86-9 Purity Grade: ACS Grade Physical Form: Granular MDL Number: MFCD00011232
  spinner