Dichloroethane -1 2-
1,2-Dichloroethane, 99+%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane Name Note: 99+% CAS: 107-06-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy ACS Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane, 99+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane, 99.8+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane Name Note: 99.8+% CAS: 107-06-2 Purity Grade: Extra Pure Assay: ≥99.80% (GC) Assay Percent Range: 99.8+% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 Purity Grade: Certified ACS Reagent Grade Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C2H4Cl2 Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: Ethylene Chloride
1,2-Dichloroethane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane Name Note: 99+% CAS: 107-06-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane, extra pure, ACROS Organics™
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 Purity Grade: Extra Pure Assay: ≥99.0% (GC) Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane, 99.5%, for electronic use, residue free, ACROS Organics™
Chemical Name or Material: 1, 2-Dichloroethane Name Note: Residue Free CAS: 107-06-2 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For electronic use Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
1,2-Dichloroethane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C2H4Cl2 Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: Ethylene Chloride
1,2-Propanediol OR, Macron Fine Chemicals™
Chemical Name or Material: 1, 2-Propanediol CAS: 57-55-6 Molecular Formula: CH3CH(OH)CH2OH Formula Weight: 76.1
1,2-Dichloroethane ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 Purity Grade: ACS Reagent Grade Linear Formula: ClCH2CH2Cl Formula Weight: 98.96
1,2-Dichloroethane, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane Name Note: For ACS analysis CAS: 107-06-2 Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: ClC2H4Cl Formula Weight: 98.96 Synonym: Ethylene Dichloride
1,2-Dichloroethane, 99.0% min., OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 2-Dichloroethane Name Note: OmniPure™ CAS: 107-06-2 Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: ClC2H4Cl Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 Synonym: Ethylene Dichloride
1,2-Dichloroethane, 99+%, for spectroscopy, laser grade, ACROS Organics™
Chemical Name or Material: 1, 2-Dichloroethane CAS: 107-06-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C2H4Cl2 Linear Formula: ClCH2CH2Cl Formula Weight: 98.96 MDL Number: MFCD00000963 Synonym: EDC
  spinner