Diglycolic Acid
Diglycolic acid, 98%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Diglycolic acid Name Note: 98% CAS: 110-99-6 Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: C4H6O5 Linear Formula: O(CH2CO2H)2 Formula Weight: 134.09 MDL Number: MFCD00004309 Synonym: 2, 2'-Oxydiacetic acid
  spinner