Dimethylformamide -N N-
N,N-Dimethylformamide, 99.5%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, for peptide synthesis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For peptide synthesis Assay: ≥99.8% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, HPLC Grade, greater-than99.7%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284
N,N-Dimethylformamide, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, anhydrous, 99.8%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Physical Form: Liquid Assay Percent Range: 99.8% Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide Name Note: DNase, RNase, Protease Free CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For molecular biology Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide Name Note: 99% CAS: 68-12-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3 H7 N O Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™ package of 4x25ML bottles, ACROS Organics™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Extra Dry Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.8 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.80% Assay Percent Range: 99.8+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 Synonym: DMF
N,N-Dimethylformamide (Certified ACS), Fisher Chemical
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: C3H7NO Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide Name Note: water <50 ppm CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, HPLC grade, >99.7%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284
N,N-Dimethylformamide, 99+%, for analysis, nonaqueous titration grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide Name Note: 99+% CAS: 68-12-2 Purity Grade: For analysis Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3 H7 N O Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8+%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide Name Note: 99.8% CAS: 68-12-2 CAS Min %: 99.8 CAS Max %: 100.0 Purity Grade: ACS Grade Assay: ≥99.80% Assay Percent Range: 99.8+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3 H7 N O Linear Formula: HCON(CH3)2 Formula Weight: 73.09 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide (Sequencing), Fisher BioReagents™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Sequencing Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H7NO Linear Formula: HCONCH3CH3 Formula Weight: 73.09 Synonym: DMF
N,N-Dimethylformamide ACS AR, Macron Fine Chemicals™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: ACS Reagent Grade Molecular Formula: HCON(CH3)2 Formula Weight: 73.09
N,N-Dimethylformamide, 99.8% (ACS Guaranteed Reagent), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide, 99.8% CAS: 68-12-2 Purity Grade: ACS Reagent Grade Formula Weight: 73.1
N, N-Dimethylformamide, GR, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Physical Form: Liquid Molecular Formula: C3H7NO Formula Weight: 73.11
N,N-Dimethylformamide, DriSolv™, EMD Millipore
Chemical Name or Material: N, N-Dimethylformamide Purity Grade: DriSolv Linear Formula: HCON(CH3)2 Formula Weight: 73.09
N,N-Dimethylformamide, 99.9% (for HPLC and Spectrophotometry), OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide, 99.9% CAS: 68-12-2 Formula Weight: 73.09
N,N-Dimethylformamide, Anhydrous, 99.8% min., DriSolv™, EMD Millipore
Chemical Name or Material: N, N-Dimethylformamide, Anhydrous CAS: 68-12-2 Physical Form: Liquid Assay Percent Range: 99.8% min. Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Synonym: Formic Acid Dimethylamide
N,N-Dimethylformamide, EMD Millipore
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Linear Formula: HCON(CH3)2 Formula Weight: 73.09
N,N-Dimethylformamide, DMF, GMP/ACS Grade, PHARMCO-AAPER
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Physical Form: Liquid Molecular Formula: C3H7NO Formula Weight: 73.09 Synonym: DMF
N,N-Dimethylformamide ChromAR, Macron™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09
N,N-Dimethylformamide ACS MP Biomedicals
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: ACS Reagent Grade Molecular Formula: C3H7NO Formula Weight: 73.09 Synonym: DMF
  spinner