Dioxane -1 4-
1,4-Dioxane, 99.5%, for analysis, stabilized, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane Name Note: Stabilized CAS: 123-91-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: For biochemistry Assay Percent Range: 99% Molecular Formula: C4H8O2 Formula Weight: 88.11 Synonym: Diox
1,4-Dioxane, 99%, Extra Dry, anhydrous, AcroSeal™, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diethylene dioxide
1,4-Dioxane, anhydrous, 99.8%, Alfa Aesar™
Chemical Name or Material: 1, 4-Dioxane, anhydrous CAS: 123-91-1 Physical Form: Liquid Assay Percent Range: 99.8% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571
1,4-Dioxane, >99%, Alfa Aesar™
Chemical Name or Material: 1, 4-Dioxane Name Note: For analysis ACS CAS: 123-91-1 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571
1,4-Dioxane, 99.8%, Extra Dry, Stabilized, Acros Organics™
Chemical Name or Material: 1,4-Dioxane CAS: 123-91-1 Molecular Formula: C4H8O Formula Weight: 88.11 MDL Number: MFCD00006571 UN Number: 1165 Merck Index: 153,328 Synonym: Diox
1,4-Dioxane, 99+%, ACS reagent, stabilized, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane, HPLC Grade, 99% min, Alfa Aesar™
Chemical Name or Material: 1, 4-Dioxane Name Note: Filtered through 0.2μ filters. CAS: 123-91-1 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571
1,4-Dioxane, HPLC Grade, 99% min, Alfa Aesar™
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571
1,4-Dioxane, 99+%, extra pure, stabilized, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 4-Dioxane Name Note: 99+% CAS: 123-91-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane (Histological), Fisher Chemical
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: Histological Grade MDL Number: MFCD00006571 Synonym: 1, 4-Diethylene Dioxide
1,4-Dioxane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 4-Dioxane Name Note: 99+% CAS: 123-91-1 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane Name Note: Over Molecular Sieves CAS: 123-91-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane, 99.8%, Extra Dry, stabilized, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 4-Dioxane Name Note: Water <50ppm CAS: 123-91-1 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.75% (GC) Assay Percent Range: 99.8% Molecular Formula: C4H8O2 Formula Weight: 88.11 Synonym: Diox
1,4-Dioxane, 99.5%, for HPLC, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥99.4% (GC) Assay Percent Range: 99.5% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diox
1,4-Dioxane (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: Certified ACS Reagent Grade Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: 1, 4-Diethylene Dioxide
Dioxane ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Dioxane CAS: 123-91-1 Purity Grade: ACS Reagent Grade Molecular Formula: C4H8O2 Formula Weight: 88.11
p-Dioxane, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Dioxane Name Note: For ACS analysis CAS: 123-91-1 Purity Grade: ACS/NF/USP Assay Percent Range: 99.0%, min. Molecular Formula: -O(CH2)2O(CH2)2- Linear Formula: -O(CH2)2O(CH2)2- Formula Weight: 88.1 Synonym: 1, 4-Dioxane
p-Dioxane, 99.9% min., OmniSolv, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Dioxane Name Note: OmniSolv™ CAS: 123-91-1 Purity Grade: ACS/NF/USP Assay Percent Range: 99.0%, min. Molecular Formula: -O(CH2)2O(CH2)2- Linear Formula: -O(CH2)2O(CH2)2- Formula Weight: 88.1 Synonym: 1, 4-Dioxane
p-Dioxane, Anhydrous, 99.9% min., DriSolv, EMD Millipore
Chemical Name or Material: p-Dioxane Name Note: SeccoSolv™, Dried CAS: 123-91-1 Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: C4H8O2 Formula Weight: 88.11 Synonym: 1, 4-Diethylene dioxide
1,4-Dioxane, 99.0%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: 1, 4-Dioxane CAS: 123-91-1 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.00% Molecular Formula: C4H8O2 Formula Weight: 88.11 MDL Number: MFCD00006571 Synonym: Diethylene dioxide
  spinner