Drinking Water Standards
Alternate Water Standard II (Metals), Ricca Chemical
Chemical Name or Material: Alternate Water Standard II (Metals) CAS: 7697-37-2 (Nitric Acid) CAS: 7732-18-5 (Water, Deionized)
Turbidity Standard, 10 NTU (Nephelometric Turbidity Units), Ricca Chemical
Chemical Name or Material: Turbidity Standard Name Note: 10 NTU (Nephelometric Turbidity Units) CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 9003-53-6 CAS Min %: 0.02 CAS Max %: 0.02 CAS: 26628-22-8 CAS Min %: 0.01 CAS Max %: 0.01 Physical Form: Liquid
Trace Metals I Standard, for Water, Ricca Chemical
Chemical Name or Material: Trace Metals I Standard CAS: 7732-18-5 CAS Min %: 88.0 CAS Max %: 92.0 CAS: 7697-37-2 CAS Min %: 8.05 CAS Max %: 8.38 CAS: 7784-27-2 CAS Min %: 0.57 CAS Max %: 0.59 CAS: 7789-02-8 CAS Min %: 0.07 CAS Max %: 0.07 CAS: 7440-62-2 CAS Min %: 0.02 CAS Max %: 0.02 CAS: 7440-66-6 CAS Min %: 0.01 CAS Max %: 0.01
Trace Metals II, Ricca Chemical
Chemical Name or Material: Trace Metals II CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 95.0 CAS: 7697-37-2 CAS Min %: 6.74 CAS Max %: 7.01 CAS: 7664-39-3 CAS Min %: 0.0024 CAS Max %: 0.0024 CAS: 1309-64-4 CAS Min %: 0.0019 CAS Max %: 0.0019 CAS: 7440-28-0 CAS Min %: 0.0018 CAS Max %: 0.0018 CAS: 7440-22-4 CAS Min %: 8.0E-4 CAS Max %: 8.0E-4
Turb Std, 4000 NTU, Stock Formazin Susp, APHA/EPA for Turb, EP Prim. Opalesc Susp, Ricca Chemical
Chemical Name or Material: Turbidity Standard Name Note: 4000 NTU, Stock Formazin Suspension
Primary Drinking Water Metals, Ricca Chemical
Chemical Name or Material: Primary Drinking Water Metals CAS: 7732-18-5 CAS Min %: 95.0 CAS Max %: 99.0 CAS: 7697-37-2 CAS Min %: 2.86 CAS Max %: 2.97 CAS: 10022-31-8 CAS Min %: 0.02 CAS Max %: 0.02 CAS: 7789-02-8 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 7439-92-1 CAS Min %: 9.0E-4 CAS Max %: 9.0E-4 CAS: 7440-22-4 CAS Min %: 8.0E-4 CAS Max %: 8.0E-4
Trace Metals III Standard, for Water, Ricca Chemical
Chemical Name or Material: Trace Metals III Standard CAS: 7732-18-5 CAS Min %: 90.0 CAS Max %: 93.0 CAS: 7697-37-2 CAS Min %: 7.93 CAS Max %: 8.26 CAS: 497-19-8 CAS Min %: 0.11 CAS Max %: 0.11 CAS: 471-34-1 CAS Min %: 0.11 CAS Max %: 0.11 CAS: 10022-31-8 CAS Min %: 0.09 CAS Max %: 0.09 CAS: 7664-39-3 CAS Min %: 0.04 CAS Max %: 0.04
Secondary Water Standard, Ricca Chemical
Chemical Name or Material: Secondary Water Standard CAS: 7732-18-5 CAS Min %: 95.0 CAS Max %: 99.0 CAS: 7697-37-2 CAS Min %: 2.89 CAS Max %: 3.01 CAS: 7440-66-6 CAS Min %: 0.04 CAS Max %: 0.04 CAS: 7440-50-8 CAS Min %: 0.01 CAS Max %: 0.01 CAS: 7439-89-6 CAS Min %: 0.0025 CAS Max %: 0.0026 CAS: 7439-96-5 CAS Min %: 4.0E-4 CAS Max %: 4.0E-4 Physical Form: Liquid
Turbidity QC Sample, Drinking Water Standard, SPEX CertiPrep™
Name Note: Yields up to 1L after dilution
Primary Drinking Water Metals without Mercury for Environmental Analysis, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Primary Drinking Water Metals without Mercury For Use With (Equipment): AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Primary Drinking Water Metals For Environmental Analysis, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Primary Drinking Water Metals For Use With (Equipment): AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Cyanitc Acid Test Solution, LaMotte™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyanitc Acid Test Solution CAS: 108-78-1 CAS: 64-19-7 CAS: 127-08-2 CAS: 7732-18-5 Physical Form: Liquid
Turbidity Standard 100 NTU (2020i / TC3000i), LaMotte™
Chemical Name or Material: Turbidity Standard Name Note: 100 NTU (2020i / TC3000i) CAS: 9003-70-7 CAS: 7732-18-5 Physical Form: Liquid
Secondary Drinking Water Metals For Environmental Analysis, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Secondary Drinking Water Metals Purity Grade: For Environmental Analysis Physical Form: Liquid
Cyanide QC Sample, Drinking Water Standard, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Name Note: Yields up to 2L after dilution
o-Phosphate Nutrients Drinking Water QC Sample, SPEX CertiPrep™
Name Note: Yields up to 2L after dilution
Residual Chlorine QC Sample, SPEX CertiPrep™
Name Note: Yields up to 2L after dilution
Drinking Water Standards without Mercury for Environmental Analysis, Set of 2, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Drinking Water Standards without Mercury Purity Grade: For Environmental Analysis Physical Form: Set Of 2 Drinking Water Pollutant Standard
Hydrazine Test Solutions, LaMotte™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hydrazine Test Solutions
Nickel Test Solutions, LaMotte™
Chemical Name or Material: Nickel Test Solutions
Nitrite Drinking Water QC Sample, SPEX CertiPrep™
Name Note: Yields up to 2L after dilution
Mercury Drinking Water QC Sample, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
Name Note: Yields up to 1L after dilution
Haloacetic Acids Mixture, Various concentrations in methyl tert-butyl ether, seven analytes, Ultra Scientific
Chemical Name or Material: Haloacetic Acids Mixture CAS: 001634-04-4 CAS Max %: 98.11 CAS: 000079-11-8 CAS Max %: 0.41 CAS: 000079-43-6 CAS Max %: 0.41 CAS: 000076-03-9 CAS Max %: 0.14 CAS: 000079-08-3 CAS Max %: 0.27 CAS: 005589-96-8 CAS Max %: 0.27
Haloacetic Acids Mixture, Various concentrations in methyl tert-butyl ether, eleven analytes, Ultra Scientific
Chemical Name or Material: Haloacetic Acids Mixture CAS: 001634-04-4 CAS Max %: 97.3 CAS: 000079-11-8 CAS Max %: 0.41 CAS: 000079-43-6 CAS Max %: 0.41 CAS: 000076-03-9 CAS Max %: 0.14 CAS: 000079-08-3 CAS Max %: 0.27 CAS: 005589-96-8 CAS Max %: 0.27
Cyanide Drinking Water/Wastewater Standard (ULTRAcheck™ Blind QC Standard)
Chemical Name or Material: Cyanide Drinking Water/Wastewater Standard Name Note: ULTRAcheck Blind QC Standard
  spinner